Skip to Content
Merck
All Photos(3)

Key Documents

NA2120

Sigma-Aldrich

GenElute Bacterial Genomic DNA Kits

sufficient for 350 purifications

Synonym(s):

Bacterial Genomic DNA, bacterial DNA prep, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.55

usage

sufficient for 350 purifications

Quality Level

technique(s)

DNA purification: suitable

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The GenElute Bacterial Genomic DNA kit provides a simple and convenient way to isolate pure genomic DNA from gram-negative bacteria. For most gram-positive bacteria, the kit must be used in conjunction with the optional lysozyme (L4919) to effectively lyse the thick peptidoglycan cell walls. A Gram-Positive Lysis Solution is provided as a diluent for preparing the lysozyme stock solutions.

The kit combines the advantages of a silica-based system with a microspin format and eliminates the need for expensive resins, alcohol precipitation, and hazardous organic compounds such as phenol and chloroform.

Application

The purified bacterial genomic DNA is ready for downstream applications such as:
  • restriction endonuclease digestions
  • PCR
  • Southern blots
  • cloning
The purified bacterial genomic DNA is ready for downstream applications such as:
  • restriction endonuclease digestions
  • PCR
  • Southern blots
  • cloning

GenElute Bacterial Genomic DNA Kits has been used:
  • to isolate DNA
  • to prepare DNA from E. coli strain
  • to extract genomic DNA

Features and Benefits

  • Starting material: Up to 1.5 mL of culture
  • Expected yield: Up to 20 μg
  • Elution volume: 400 μl
  • Time required: 70 - 120 min
  • A260/A280 ratio: 1.6 - 1.9
  • No phenol, chloroform, or ethanol precipitation required

Principle

The bacteria are lysed in a chaotropic salt-containing solution to ensure the thorough denaturation of macromolecules. The addition of ethanol causes the DNA to bind when the lysate is spun through a silica membrane into a microcentrifuge tube. After washing to remove the contaminants, the DNA is eluted in 200 μL of a Tris-EDTA solution.

The expected yield of genomic DNA will vary depending on the cell density of the bacterial culture and the bacterial species and strain used. DNA purified with the GenElute kit has an A260/A280 ratio between 1.6 and 1.9 and can be up to 50 kb in length.

Other Notes

For additional information, please see www.sigma-aldrich.com/genomicdna.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • W0263Wash Solution 1SDS

  • C2112Column Preparation SolutionSDS

  • P2308Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biologySDS

  • R6148RNase A solutionSDS

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margaret T Ho et al.
Genome announcements, 5(24) (2017-06-18)
We report here the complete assembled genome sequence of Acinetobacter calcoaceticus CA16, which is capable of utilizing diesel and lignin as a sole carbon source. CA16 contains a 4,110,074-bp chromosome and a 5,920-bp plasmid. The assembled sequences will help elucidate
Urinary tract Infection Escherichia coli is related to the environmental Escherichia coli in their DNA barcoding and antibiotic resistance patterns in Grenada
Farmer K, et al.
Advances in Microbiology, 6(01), 33-33 (2016)
Nora Vázquez-Laslop et al.
Journal of bacteriology, 188(10), 3494-3497 (2006-05-05)
Bacterial populations contain persisters, cells which survive exposure to bactericidal antibiotics and other lethal factors. Persisters do not have a genetic resistance mechanism, and their means to tolerate killing remain unknown. In exponentially growing populations of Escherichia coli the frequency
Ikrema Hassan et al.
Genome announcements, 4(1) (2016-01-16)
Here we report the complete genomic sequence of the bacterium Arthrobacter sp. strain LS16, consisting of a single circular chromosome of 3.85 Mb with no identified plasmid. Data contained within will facilitate future genetic modification and engineering of the Arthrobacter sp.
Modulation of Global Transcriptional Regulatory Networks as a Strategy for Increasing Kanamycin Resistance of the Translational Elongation Factor-G Mutants in Escherichia coli
Mogre A, et al.
G3 (Bethesda, Md.), g3-300284 (2017)

Protocols

GenElute™ Bacterial Genomic DNA Kit protocol describes a simple and convenient way for the isolation of pure genomic DNA from bacteria.

GenElute™ Bacterial Genomic DNA Kit protocol describes a simple and convenient way for the isolation of pure genomic DNA from bacteria.

GenElute™ Bacterial Genomic DNA Kit protocol describes a simple and convenient way for the isolation of pure genomic DNA from bacteria.

GenElute™ Bacterial Genomic DNA Kit protocol describes a simple and convenient way for the isolation of pure genomic DNA from bacteria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service