NA0800
GenElute™ HP Select Plasmid Gigaprep Kit
sufficient for 5 preparations
Synonym(s):
HP gigaprep kit, endo free gigaprep kit, endotoxin free giga kit, endotoxin free plasmid kit, fast gigaprep kit, fast plasmid isolation, plasmid extraction kit, plasmid isolation kit, plasmid purification kit, ultrafast plasmid isolation, Gen Elute, GenElute™ HP Plasmid Kit
About This Item
Recommended Products
usage
sufficient for 5 preparations
Quality Level
greener alternative product characteristics
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
DNA extraction: suitable
greener alternative category
storage temp.
15-25°C
General description
An overnight recombinant E. coli culture is harvested by centrifugation and subjected to a modified alkaline-SDS lysis. The lysate is cleared by filtration followed by the addition of a binding solution optimized for endotoxin-free plasmid preparations. The plasmid DNA is then captured onto a silica membrane in the presence of high salts while endotoxins are prevented from adsorbing to the membrane. Contaminants are removed with three wash steps. Finally, the bound DNA is eluted in endotoxin-free water.
The DNA is ready for immediate use in applications such as restriction digest, cloning, sequencing, PCR, and transfection of endotoxin-sensitive cell lines.
Application
Features and Benefits
- Up to 15 mg with endotoxin levels of ≤ 0.1 EU/μg
- Only 90 minutes from pelleted cells to purified plasmid
- Vacuum format with no ethanol precipitation
- Can purify low-, medium-, and high-copy plasmid DNA
Other Notes
Legal Information
Kit Components Also Available Separately
- G3671GenElute™ HP Pro Gigaprep Binding ColumnSDS
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3
Target Organs
Central nervous system, Lungs
Storage Class Code
3 - Flammable liquids
Flash Point(F)
77.0 °F - closed cup
Flash Point(C)
25 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service