N8159
Albumin, Nitrated from bovine serum
lyophilized powder
Synonym(s):
Nitrated BSA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
bovine
Quality Level
form
lyophilized powder
technique(s)
immunoblotting: suitable
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
bovine ... ALB(280717)
General description
Albumin, Nitrated (Nitrated BSA), is prepared by chemical modification of bovine serum albumin (BSA) by peroxynitrite. Peroxynitrite (ONOO-), a potent oxidant is an important member of the family of reactive oxygen and nitrogen species.
Application
Positive control in immunoblotting assays for Anti-Nitrotyrosine (Sigma Product Number N 0409).
Biochem/physiol Actions
Peroxynitrite promotes the nitration of tyrosine residues in proteins linked, oxidation of redox metal centers, DNA, lipids and nitrosation of cysteine residues in proteins, suggesting that peroxynitrite plays a major role in oxidative cellular damage. Peroxynitrite has been implicated in the pathogenesis of several inflammatory, infectious and degenerative human disease. These include neurological conditions such as Alzheimer′s disease, amyotrophic lateral sclerosis (ALS), the pathogenesis of asthma, atherosclerosis, and a variety of conditions precipitated by endothelial injury.
Physical form
Lyophilized from 0.01 M phosphate buffered saline, pH 7.4
Preparation Note
Bovine serum albumin (BSA) nitrated with peroxynitrite.
Analysis Note
Does not contain sodium azide
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Intracellular Redox Stress Caused by Hexavalent Chromium is Selective for Proteins that Have Key Roles in Cell Survival and Thiol Redox Control
Toxicology (2011)
Inhibitory effect of isoflavones on peroxynitrite-mediated low-density lipoprotein oxidation
Bioscience, Biotechnology, and Biochemistry, 66(1), 22-28 (2002)
Nitric oxide and peroxynitrite in health and disease
Physiological Reviews, 87(1), 315-424 (2007)
Antioxidants (Basel, Switzerland), 10(1) (2021-01-13)
Selenium (Se) enrichment of Stevia rebaudiana Bertoni can serve a dual purpose, on the one hand to increase plant biomass and stress tolerance and on the other hand to produce Se fortified plant-based food. Foliar Se spraying (0, 6, 8
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service