Skip to Content
Merck
All Photos(4)

Key Documents

N7251

Sigma-Aldrich

4-Nitrocatechol sulfate dipotassium salt

crystalline

Synonym(s):

2-Hydroxy-5-nitrophenyl sulfate, 2-Hydroxy-5-nitrophenyl sulfate dipotassium salt monohydrate, p-Nitrocatechol sulfate dipotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3NO7SK2
CAS Number:
Molecular Weight:
311.35
Beilstein:
3805018
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

crystalline

color

yellow

storage temp.

−20°C

SMILES string

[K+].[K+].[O-]c1ccc(cc1OS([O-])(=O)=O)[N+]([O-])=O

InChI

1S/C6H5NO7S.2K/c8-5-2-1-4(7(9)10)3-6(5)14-15(11,12)13;;/h1-3,8H,(H,11,12,13);;/q;2*+1/p-2

InChI key

CKWWBDCAYRJAJB-UHFFFAOYSA-L

General description

4-Nitrocatechol sulfate is an aromatic sulfate.

Application

4-Nitrocatechol sulfate dipotassium salt has been used as a substrate in mucopolysaccharidoses VI (MPS VI) analysis. It has also been used to measure the activity of glycosaminoglycan (GAG)-degrading enzymes-arylsulfatase B, and exoglycosidases.

Substrates

Chromogenic sulfatase substrate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The convulsive effects of dopamine sulfate conjugates in rat brain.
N T Buu et al.
Life sciences, 29(22), 2311-2316 (1981-11-30)
Sandra Del Carmen Mendoza-Ruvalcaba et al.
Genetics and molecular biology, 43(1), e20180347-e20180347 (2020-02-28)
Mucopolysaccharidoses (MPS) are a group of genetic disorders, each resulting from the deficiency of one of the lysosomal enzymes that catabolizes mucopolysaccharides. For the accurate diagnosis of the disease, the quantification of a specific enzymatic activity is needed. In the
Camila M Adade et al.
Micron (Oxford, England : 1993), 38(3), 252-256 (2006-07-25)
Lysosomes of trypanosomatid protozoa are poorly known. In this work we have cytochemically detected the lysosomal enzyme aryl sulphatase in the trypanosomatids Trypanosoma cruzi and Crithidia fasciculata, by using p-nitrocatecholsulphate as substrate. Positive reaction was located exclusively inside membrane-bound cytoplasmic
Jin-Peng Sun et al.
The Journal of biological chemistry, 278(35), 33392-33399 (2003-06-18)
The pathogenic bacteria Yersinia are causative agents in human diseases ranging from gastrointestinal syndromes to bubonic plague. There is increasing risk of misuse of infectious agents, such as Yersinia pestis, as weapons of terror as well as instruments of warfare
D B Thompson et al.
Experientia, 42(2), 150-151 (1986-02-15)
Rodent and bovine arylsulfatase B hydrolyze 4-methylumbelliferyl sulfate (4MUS) 10- to 30-fold more efficiently than arylsulfatase A. Therefore, 4MUS grossly underestimates arylsulfatase A activity in the presence of excess arylsulfatase B.

Articles

Drug conjugate analysis and the enzymatic hydrolysis of glucuronides

Protocols

Enzymatic Assay of Sulfatase (EC 3.1.6.1.)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service