Skip to Content
Merck
All Photos(3)

Key Documents

N3915

Sigma-Aldrich

Notoginsenoside R1

≥98% (HPLC)

Synonym(s):

(3β,6α,12β)-20-(β-D-Glucopyranosyloxy)-3,12-dihydroxydammar-24-en-6-yl 2-O-β-D-xylopyranosyl-β-D-glucopyranoside, Sanchinoside R1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H80O18
CAS Number:
Molecular Weight:
933.13
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

biological source

Panax notoginseng (Burk.) F.H.Chen

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

H2O: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

C\C(C)=C\CC[C@](C)(O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H]2CC[C@]3(C)[C@@H]2[C@H](O)C[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5[C@H](C[C@@]34C)O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O[C@@H]7OC[C@@H](O)[C@H](O)[C@H]7O

InChI

1S/C47H80O18/c1-21(2)10-9-13-47(8,65-41-37(59)34(56)32(54)26(18-48)62-41)22-11-15-45(6)30(22)23(50)16-28-44(5)14-12-29(52)43(3,4)39(44)25(17-46(28,45)7)61-42-38(35(57)33(55)27(19-49)63-42)64-40-36(58)31(53)24(51)20-60-40/h10,22-42,48-59H,9,11-20H2,1-8H3/t22-,23+,24+,25-,26+,27+,28+,29-,30-,31-,32+,33+,34-,35-,36+,37+,38+,39-,40-,41-,42+,44+,45+,46+,47-/m0/s1

InChI key

LLPWNQMSUYAGQI-OOSPGMBYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Notoginsenoside R1 has been shown to exhibit antooxidant, anti-inflammatory, antiapoptotic, and immune-stimulatory properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jian-Qing Ruan et al.
Journal of agricultural and food chemistry, 58(9), 5770-5776 (2010-04-22)
As a main and characteristic constituent in Radix notoginseng, the fate of notoginsenoside R1 (NGR1) in human is largely unknown. The present study investigated, for the first time, NGR1 metabolism by human intestinal bacteria and liver subcellular fractions, and permeability
Shan Cong et al.
International immunopharmacology, 68, 137-144 (2019-01-10)
Pneumonia is a common lung disease in children with high fatality rate. Notoginsenoside R1 (NGR1) is the main active component extracted from the roots of Panax notoginseng (Burk.) F.H. Chen (Araliaceae). Here, we carefully explored the potential anti-inflammatory and protective
T T X Dong et al.
Phytotherapy research : PTR, 19(8), 684-688 (2005-09-24)
The root of Panax notoginseng (Radix Notoginseng, Sanqi) is a commonly used traditional Chinese medicine, which is mainly cultivated in Wenshan of Yunnan China. The identified active constituents in Radix Notoginseng include saponin, ssavonoid and polysaccharide; however, the levels of
Kai Sun et al.
Life sciences, 81(6), 509-518 (2007-07-28)
Ginsenoside Rb1 (Rb1), ginsenoside Rg1 (Rg1), and notoginsenoside R1 (R1) are major active components of Panax notoginseng, a Chinese herb that is widely used in traditional Chinese medicine to enhance blood circulation and dissipate blood stasis. To evaluate the effect
Kouichi Yoshizaki et al.
Chemical & pharmaceutical bulletin, 60(3), 354-362 (2012-03-03)
Seven new dammarane-type triterpenoid saponins, chikusetsusaponin FK1 (1), chikusetsusaponin FK2 (2), chikusetsusaponin FK3 (3), chikusetsusaponin FK4 (4), chikusetsusaponin FK5 (5), chikusetsusaponin FK6 (6), and chikusetsusaponin FK7 (7), and eleven known triterpenoid saponins, ginsenoside Rb3 (9), ginsenoside Rc (10), chikusetsusaponin VI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service