Skip to Content
Merck
All Photos(3)

Key Documents

N17001

Sigma-Aldrich

Noggin human

recombinant, expressed in HEK 293 cells, suitable for cell culture

Synonym(s):

Noggin human

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in HEK 293 cells

Quality Level

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≤10 ng/mL ED50

mol wt

23 kDa (The protein migrates as a 25 kDa band on SDS-PAGE due to glycosylation)

technique(s)

cell culture | mammalian: suitable

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Recombinant human Noggin is expressed in human 293 cells as a glycoprotein with a calculated molecular mass of 23 kDa. This protein is manufactured in human cells using an all-human production system, with no serum. The human cells expression system allows human-like glycosylation and folding, and often supports better stability of the protein in culture.

Biochem/physiol Actions

Noggin is a secreted protein that inhibits the binding of bone morphogenetic proteins (BMPs) to their cognate receptor. It is a 232 amino acid-secreted glycosylated protein, which forms covalently linked homodimers and has high affinity for BMP4. hESC cultured with noggin (in medium or incorporated into extracellular matrix) form denser colonies compared to normal hESC cultures, suggesting that the presence of noggin promotes better growth. Noggin can be incorporated as a medium supplement for maintaining stem cells in a pluripotent state, for short-term culture experiments. Noggin does not trigger differentiation towards a neuronal lineage. Furthermore, when incorporated into extracellular matrix, noggin prevented spontaneous differentiation during the time period examined. In a surgically induced knee osteoarthritis model in mice, expression of noggin mRNA was lost from the articular cartilage, which correlated with loss of BMP2/4 and pSMAD1/5/8, an indicator of active BMP signaling.

Sequence

QHYLHIRPAPSDNLPLVDLIEHPDPIFDPKEKDLNETLLRSLLGGHYDPGFMATSPPEDRPGGGGGAAGGAEDLAELDQLLRQRPSGAMPSEIKGLEFSEGLAQGKKQRLSKKLRRKLQMWLWSQTFCPVLYAWNDLGSRFWPRYVKVGSCFSKRSCSVPEGMVCKPSKSVHLTVLRWRCQRRGGQRCGWIPIQYPIISECKCSC

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Analysis Note

The biological activity of recombinant human noggin was tested in culture by measuring its ability to inhibit BMP4-induced alkaline phosphatase production by ATDC5 cells (human erythroleukemic indicator cell line).
The EC50 is defined as the effective concentration of growth factor that elicits a 50% decrease in alkaline phosphatase secretion in a cell based bioassay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The expression patterns of gremlin 1 and noggin in normal adult and tumor tissues.
Laurilla R, et al.
International Journal of Clinical and Experimental Pathology, 6(7), 1400-1408 (2013)
Wenjing Xiao et al.
Autophagy, 18(11), 2615-2635 (2022-03-08)
Macroautophagy/autophagy is a conserved cellular process associated with tumorigenesis and aggressiveness, while mechanisms regulating expression of autophagic machinery genes in cancers still remain elusive. Herein, we identified E2F4 (E2F transcription factor 4) as a novel transcriptional activator of cytoprotective autophagy

Articles

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service