Skip to Content
Merck
All Photos(1)

Key Documents

N1638

Sigma-Aldrich

Nystatin Suspension

suspension, non-animal origin, suitable for cell culture, BioReagent

Synonym(s):

Fungicidin, Mycostatin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.76

Product Name

Nystatin Suspension, suspension, 10,000 unit/mL in DPBS, BioReagent, suitable for cell culture

product line

BioReagent

Quality Level

form

suspension

concentration

10,000 unit/mL in DPBS

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

yellow

antibiotic activity spectrum

fungi
yeast

Mode of action

cell membrane | interferes

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@@H]1OC(=O)C[C@H](O)C[C@H](O)C[C@H](O)CC[C@@H](O)[C@H](O)C[C@]2(O)C[C@H](O)[C@H]([C@H](C[C@@H](O[C@@H]3O[C@H](C)[C@@H](O)[C@H](N)[C@@H]3O)\C=C\C=C\C=C\C=C\CC\C=C\C=C\[C@H](C)[C@@H](O)[C@@H]1C)O2)C(O)=O

InChI

1S/C47H75NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-36(53)35(52)20-19-31(49)21-32(50)22-33(51)23-39(55)62-29(3)28(2)42(27)56/h5-6,8,10-18,27-38,40-44,46,49-54,56-58,61H,7,9,19-26,48H2,1-4H3,(H,59,60)/b6-5+,10-8+,13-11+,14-12+,17-15+,18-16+/t27-,28+,29-,30+,31+,32+,33+,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

VQOXZBDYSJBXMA-QEKUPDCNSA-N

General description

Chemical structure: polyene

Application

Nystatin suspension has been used:
  • to inhibit caveolin-mediated endocytosis
  • as a component of Eagle′s minimum essential medium (EMEM)
  • to enumerate lactic acid bacteria (LAB)
  • in preventing cell culture contamination and inducing interleukin (IL)-1, IL-8, and tumor necrosis factor a secretion in toll-like receptor 2 (TLR2)-expressing THP1 cells.
This product is recommended for use in cell culture applications at 24 mL/L.

Biochem/physiol Actions

Nystatin is a polyene, broad-spectrum antibiotic, which modifies the microstructure of lipid rafts. It can sequester cholesterol present on the cell membrane of eukaryotes. Nystatin regulates immune responses through cytokine signaling, lymphocyte activation and pathogen recognition.

Caution

This product is stable at 37°C for three days.

Preparation Note

It is 0.1μ filtered and cell culture tested.

Other Notes

Nystatin suspension available in 20 mL and 100 mL pack size.
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A method to maintain mammalian cells for days alive at 4 C
Moce-Llivina L and Jofre J
Cytotechnology, 46(1), 57-61 (2004)
Liu He et al.
eLife, 9 (2020-04-16)
The development of a polarized neuron relies on the selective transport of proteins to axons and dendrites. Although it is well known that the microtubule cytoskeleton has a central role in establishing neuronal polarity, how its specific organization is established
Ian T Stancil et al.
Nature communications, 12(1), 4566-4566 (2021-07-29)
The airway epithelium serves as the interface between the host and external environment. In many chronic lung diseases, the airway is the site of substantial remodeling after injury. While, idiopathic pulmonary fibrosis (IPF) has traditionally been considered a disease of
Conservation characteristics of corn ears and stover ensiled with the addition of Lactobacillus plantarum MTD-1, Lactobacillus plantarum 30114, or Lactobacillus buchneri 11A44
Lynch JP, et al.
Journal of Dairy Science, 95(4), 2070-2080 (2012)
Nystatin enhances the immune response against Candida albicans and protects the ultrastructure of the vaginal epithelium in a rat model of vulvovaginal candidiasis
Zhang X, et al.
BMC Microbiology, 18(1), 166-166 (2018)

Articles

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service