Skip to Content
Merck
All Photos(1)

Key Documents

N1038

Sigma-Aldrich

Anti-Nup98 antibody, Rat monoclonal

clone 2H10, purified from hybridoma cell culture

Synonym(s):

Anti-Nucleoporin, 98-kD

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2H10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~98 kDa

species reactivity

rat, human

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
western blot: 0.5-1 μg/mL using HeLa nuclear extract

isotype

IgG2cκ

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUP98(4928)
rat ... Nup98(81738)

General description

Monoclonal Anti-Nup98 (rat IgG2c- isotype) is derived from the hybridoma 2H10 produced by the fusion of mouse myeloma cells (SP2 cells) and splenocytes from WKY/NCrj rat immunized with a recombinant fragment of human Nup98. Nup96 and Nup98 are the nucleoporins that are formed by the cleavage of 186 kDa precursor protein.
Nucleoporin-98kDa (Nup98) is part of the nuclear pore complex. It possesses two domains and the gene encoding it is localized on human chromosome 11.

Application

Anti-Nup98 antibody, Rat monoclonal may be used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunocytochemistry

Biochem/physiol Actions

Nucleoporin-98kDa (Nup98) is involved in the translocation of RNA and proteins across the nucleus. It is also a regulator of chromosomal segregation and gene expression. The protein also binds to other nucleoporins and is associated with acute myelogenous leukemia.
Nup98 functions as a docking protein for cytosol mediated docking of import substrates. Securin protein damage caused by the anaphase-promoting complex (APC) during mitosis, is regulated by the nucleocytoplasmic transport factors Rae1 and Nup98. In mice, collective haploinsufficiency of Rae1 and Nup98 gene causes premature separation of sister chromatids and severe aneuploidy. In addition, it also leads to untimely degradation of securin. Chromosome rearrangement between the HOXA9 (homeobox A9) gene and the nucleoporin gene Nup98 has been observed in some human myeloid leukemia patients.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Céline Ricour et al.
Journal of virology, 83(21), 11223-11232 (2009-08-28)
The leader protein of cardioviruses, Theiler's murine encephalomyelitis virus (TMEV) and encephalomyocarditis virus (EMCV), is a multifunctional protein known to antagonize type I interferon expression and to interfere with nucleocytoplasmic trafficking of host proteins and mRNA. This protein plays an
Valentina Salsi et al.
Cancer research, 74(4), 1079-1090 (2013-12-29)
NUP98 is a recurrent fusion partner in chromosome translocations that cause acute myelogenous leukemia. NUP98, a nucleoporin, and its interaction partner Rae1, have been implicated in the control of chromosome segregation, but their mechanistic contributions to tumorigenesis have been unclear.
NUP98 gene fusions and hematopoietic malignancies: common themes and new biologic insights
Gough SM, et al.
Blood, 118(24), 6247-6257 (2011)
Takaomi Fukuhara et al.
Hybridoma (2005), 24(5), 244-247 (2005-10-18)
Nup98 is a component of nuclear pore complexes, which are large protein assemblies embedded in the nuclear envelope. Previous studies have shown that Nup98 interacts with several transport factors and plays a critical part in nuclear trafficking. However, the mechanism
Masaaki Iwamoto et al.
Genes to cells : devoted to molecular & cellular mechanisms, 15(7), 661-669 (2010-06-16)
The nucleoporin Nup98 is an essential component of the nuclear pore complex. This peripheral nucleoporin with its Gly-Leu-Phe-Gly (GLFG) repeat domain contributes to nuclear-cytoplasmic trafficking, including mRNA export. In addition, accumulating studies indicate that Nup98 plays roles in several important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service