Skip to Content
Merck
All Photos(1)

Key Documents

N0165

Sigma-Aldrich

Nisoldipine

≥98% (HPLC)

Synonym(s):

(±)-Isobutyl methyl 1,4-dihydro-2,6-dimethyl-4-(o-nitrophenyl)-3,5-pyridinedicarboxylate, 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic acid methyl 2-methylpropyl ester, Sular

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O6
CAS Number:
Molecular Weight:
388.41
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: >10 mg/mL

originator

Bayer

storage temp.

room temp

SMILES string

COC(=O)C1=C(C)NC(C)=C(C1c2ccccc2[N+]([O-])=O)C(=O)OCC(C)C

InChI

1S/C20H24N2O6/c1-11(2)10-28-20(24)17-13(4)21-12(3)16(19(23)27-5)18(17)14-8-6-7-9-15(14)22(25)26/h6-9,11,18,21H,10H2,1-5H3

InChI key

VKQFCGNPDRICFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nisoldipine has been used:
  • as a hit-antagonist to study its effects on oxytocin (OT)-induced myometrial Ca2+-mobilization and its potency to dampen uterine myometrial contractions [1]
  • as a calcium channel blocker to study its effects on cell viability in a human hepatic cancer HepG2 and rat renal epithelial NRK52E cell line [2]
  • as a component of the external IK1 recording solution to study the action potential in human pluripotent stem cell−derived cardiomyocytes (hiPS-CMs)[3]

Biochem/physiol Actions

L-type (CaV1.2) calcium channel blocker; dihydropyridine-type calcium channel blocker with selectivity for smooth muscle (CaV1.2b) over cardiac muscle (CaV1.2a). Arterial vasodilator and antihypertensive agent.

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer L Herington et al.
PloS one, 10(11), e0143243-e0143243 (2015-11-26)
The uterine myometrium (UT-myo) is a therapeutic target for preterm labor, labor induction, and postpartum hemorrhage. Stimulation of intracellular Ca2+-release in UT-myo cells by oxytocin is a final pathway controlling myometrial contractions. The goal of this study was to develop
Xue-yan Chen et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 43(10), 1011-1018 (2009-01-09)
Effect of new calcium antagonist m-nisoldipine (m-Nis) on MCT-induced PH in rats and its mechanisms were investigated. Rats were injected with a single dose (60 mg x kg(-1)) of MCT subcutaneously to induce PH. Pulmonary haemodynamic measurement and lung tissue
Piero Ruggenenti et al.
Nature reviews. Nephrology, 6(6), 319-330 (2010-05-05)
Angiotensin II and other components of the renin-angiotensin-aldosterone system (RAAS) have a central role in the pathogenesis and progression of diabetic renal disease. A study in patients with type 1 diabetes and overt nephropathy found that RAAS inhibition with angiotensin-converting-enzyme
Tsuyoshi Shiga et al.
Heart and vessels, 22(6), 404-409 (2007-11-29)
The aim of this study was to investigate whether cilnidipine, an N- and L-type calcium channel blocker, and nisoldipine, an L-type calcium channel blocker, have different effects on sympathetic activity, using an identical group of healthy male subjects. Eight healthy
Xueyan Chen et al.
European journal of pharmacology, 686(1-3), 32-40 (2012-05-12)
The excessive proliferation of pulmonary artery smooth muscle cells (PASMCs) plays a critical role in the development of pulmonary arterial hypertension. Recent studies indicate that Ca(2+) and reactive oxygen species are critically involved in the process of smooth muscle cell

Articles

We offers many products related to calcium channels for your research needs.

We offers many products related to calcium channels for your research needs.

We offers many products related to calcium channels for your research needs.

We offers many products related to calcium channels for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service