Skip to Content
Merck
All Photos(1)

Key Documents

MSST0031

Sigma-Aldrich

SILuProt MAPT Microtubule-associated protein tau-441 human

recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C and 15N-labeled

Synonym(s):

Isotopically labeled MAPT LC/MS standard, Isotopically labeled human Microtubule-associated protein tau-441, Neurofibrillary tangle protein human, Paired helical filament-tau (PHF-tau), Pairedhelicalfilament-tau(PHF-tau), Tau-F

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≥98 Heavy amino acids incorporation efficiency by MS

technique(s)

mass spectrometry (MS): suitable

suitability

suitable for mass spectrometry (standard)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... MAPT(4137)

General description

Tau-441 is a member of the Tau family of proteins. Tau proteins are mainly expressed in the neurons of the central nervous system where they exert a role in stabilizing microtubules, key components of axonal transport, as well as in signal transduction.Tau proteins are subject to phosphorylation and this phenomenon regulates the association of the Tau protein with the microtubules. Deposits of Alzheimer′s disease AD-associated proteins, such as hyperphosphorylated Tau, as well as other shared misfolded proteins, such as, β-amyloid precursor protein (βAPP), ubiquitin, and various chaperones and protein kinases are thought to play a pathologic role in the cognitive decline and muscular failure.3 Malfunctioning of Tau proteins is associated with microtubules disintegration and collapsing of the neuronal transport system. Among other diseases, Tau forms in cerebrospinal fluid are considered a reliable biomarker for progressive supranuclear palsy, where the levels of Tau forms ratio were significantly reduced.

Immunogen

MAEPRQEFEVMEDHAGTYGLGDRKDQGGYTMHQDQEGDTDAGLKESPLQTPTEDGSEEPGSETSDAKSTPTAEDVTAPLVDEGAPGKQAAAQPHTEIPEGTTAEEAGIGDTPSLEDEAAGHVTQARMVSKSKDGTGSDDKKAKGADGKTKIATPRGAAPPGQKGQANATRIPAKTPPAPKTPPSSGEPPKSGDRSGYSSPGSPGTPGSRSRTPSLPTPPTREPKKVAVVRTPPKSPSSAKSRLQTAPVPMPDLKNVKSKIGSTENLKHQPGGGKVQIINKKLDLSNVQSKCGSKDNIKHVPGGGSVQIVYKPVDLSKVTSKCGSLGNIHHKPGGGQVEVKSEKLDFKDRVQSKIGSLDNITHVPGGGNKKIETHKLTFRENAKAKTDHGAEIVYKSPVVSGDTSPRHLSNVSSTGSIDMVDSPQLATLADEVSASLAKQGLDRIRGRKLGPFEGKPIPNPLLGLDSTRTGHHHHHHHHGGQ

Biochem/physiol Actions

SILuProt MAPT is a recombinant, stable isotope-labeled human MAPT which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of MAPT in mass-spectrometry. SILu Prot MAPT is a protein of 481 amino acids (including C-terminal V5 and polyhistidine tags) with a calculated molecular mass of 36 kDa.

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service