MAK520
Monoamine Oxidase (MAO) Inhibitor Screening Kit
sufficient for 100 fluorometric tests
Synonym(s):
MAO-A Inhibitor Detection Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
Quality Level
application(s)
pharmaceutical
detection method
fluorometric
relevant disease(s)
Alzheimer′s disease; Parkinson′s disease; depression; migraine disorder; schizophrenia; substance abuse; neurodegenerative disorders; neuropsychiatric diseases; neurological disorders
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
Monoamine oxidases (MAO) are a family of mitochondrial enzymes that catalyze the oxidative deamination of monoamines. Two isoforms of MAO exist, MAO-A and MAO-B, with different inhibitor selectivity and tissue distribution. MAO dysfunction is thought to be responsible for a number of neurological disorders. Unusually high or low levels of MAOs in the body have been associated with depression, schizophrenia, substance abuse, attention deficit disorder, migraines, and irregular sexual maturation. MAO inhibitors are one of the major classes of drug prescribed for the treatment of depression, Parkinson′s, and Alzheimer′s diseases.
Features and Benefits
- Compatible with high-throughput handling systems.
Suitability
The kit is suitable for inhibitor screening and evaluation of MAO inhibitors.
Principle
The Monoamine Oxidase (MAO) Inhibitor Screening Kit provides a convenient fluorometric means to screen for MAO enzyme inhibitors. In the assay, MAO reacts with p-tyramine, a substrate for both MAO-A and MAO-B, resulting in the formation of H2O2, which is determined by a fluorometric method at λEx = 530 nm/λEm = 585 nm.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service