MAK478
α-Amylase Activity Assay Kit
sufficient for 100 colorimetric tests
Synonym(s):
Assay Kit for Amylase Activity
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
NACRES:
NA.84
Recommended Products
Quality Level
application(s)
agriculture
food and beverages
detection method
colorimetric
relevant disease(s)
endocrinological disorders; gastrointestinal diseases; genitourinary disorders
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
Amylase belongs to the family of glycoside hydrolase enzymes that break down starch into glucose molecules by acting on α-1,4- glycosidic bonds. The α-amylases cleave at random locations on the starch chain, ultimately yielding maltotriose and maltose, glucose and "limit dextrin" from amylose and amylopectin. In mammals, α-amylase is a major digestive enzyme. Increased enzyme levels in humans are associated with salivary trauma, mumps due to inflammation of the salivary glands, pancreatitis, and renal failure.
Features and Benefits
- The linear detection range of the kit is 0.3 to 50 U/L α-amylase.
Suitability
The kit is suitable for the determination of α-amylase activity in blood, saliva, urine, grains, and other agricultural samples.
Principle
Simple, direct, and automation-ready procedures for measuring amylase activity are very desirable. The α-Amylase Activity Assay Kit method involves two steps. First, α -amylase in the sample hydrolyses starch and the product is rapidly converted to glucose by α-glucosidase and hydrogen peroxide by glucose oxidase. Next, the hydrogen peroxide concentration is determined with a colorimetric reagent at 585 nm.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service