Skip to Content
Merck
All Photos(2)

Key Documents

MAK108

Sigma-Aldrich

PLTP Activity Assay Kit

Supplied by Roar Biomedical, Inc.

Synonym(s):

Phospholipid transfer protein activity assay kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84
Pricing and availability is not currently available.

usage

sufficient for 100 fluorometric tests

application(s)

pharmaceutical

detection method

fluorometric

relevant disease(s)

cardiovascular diseases

storage temp.

2-8°C

Gene Information

human ... PLTP(5360)

General description

Phospholipid transfer protein (PLTP) is a protein present in normal human plasma. PLTP transfers phospholipids among lipoproteins in plasma.

Suitability

Suitable to determine Phospholipid transfer protein (PLTP) activity in plasma or serum (fresh or frozen) as well as other sources of PLTP.

Principle

The PLTP Activity Assay Kit includes proprietary substrates to detect PLTP mediated transfer of fluorescent substrate. Transfer activity results in increased fluorescent emission intensity (λEx = 465 nm/λEm = 535 nm) from the assay.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan J Henderson et al.
Drug development and industrial pharmacy, 35(9), 1139-1146 (2009-04-22)
To investigate how different formulations of Amphotericin-B (Amp-B) affect the activity of phospholipid transfer protein (PLTP) when incubated with hyperlipidemic and normolipidemic plasma at physiological temperature (37 degrees C). Six hyperlipidemic and six normolipidemic plasma samples were collected and tested
Axel Schlitt et al.
American journal of nephrology, 27(2), 138-143 (2007-02-20)
Phospholipid transfer protein (PLTP) is mainly involved in high-density lipoprotein (HDL) metabolism. The role of PLTP in atherogenesis is still controversial. We aimed to investigate PLTP activity in hemodialysis (HD) patients, a population which has an increased risk for the
Marta Marchesi et al.
British journal of pharmacology, 164(5), 1460-1468 (2011-04-14)
Besides a significant reduction of low-density lipoprotein (LDL) cholesterol, statins moderately increase high-density lipoprotein (HDL) levels. In vitro studies have indicated that this effect may be the result of an increased expression of apolipoprotein (apo)A-I, the main protein component of
Sander J Robins et al.
Atherosclerosis, 228(1), 230-236 (2013-03-13)
Cholesteryl ester transfer protein (CETP) and phospholipid transfer protein (PLTP) are two genetically-related plasma proteins involved in the exchange of cholesteryl esters and phospholipids between high-density lipoproteins (HDL) and other lipoproteins. Although low CETP and high PLTP activity both result
Nilesh Patankar et al.
Pharmaceutical research, 23(5), 1020-1024 (2006-05-23)
The purpose of this study was to investigate the role of phospholipid transfer protein (PLTP) on the plasma distribution of amphotericin B (AmpB) following incubation with different AmpB formulations in human plasmas with varying lipid profiles. In a first set

Questions

1–2 of 2 Questions  
  1. Are products MAK107 and MAK108 compatible with mouse serum?

    1 answer
    1. Yes, but please be aware of possible sample hemolysis. Depending on the method of mouse sample collection, hemolysis may be common, which can interfere with fluorescence emission via color quenching.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service