Skip to Content
Merck
All Photos(1)

Key Documents

M8316

Sigma-Aldrich

Anti-MRP2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ABCC2, Anti-Multidrug Resistance Associated Protein 2, Anti-cMOAT, Anti-cMRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~190 kDa

species reactivity

rat, human

technique(s)

indirect immunofluorescence: 1:100 using paraformaldehyde-fixed HepG2 cells
indirect immunofluorescence: 1:100 using rat liver frozen sections
western blot: 1:1,000 using whole extract of HepG2 human hepatoblastoma cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCC2(1244)
rat ... Abcc2(25303)

Related Categories

General description

Multidrug Resistance-associated Protein 2 (MRP2), also called as canalicular Multispecific Organic Anion Transporter (cMOAT), is a member of the CFTR/MRP (ABCC) subfamily of the large ATP-Binding Cassette (ABC) transporter family of transmembrane proteins. MRP2 is normally expressed in the liver, gallbladder, kidney proximal tubules, placenta, duodenum and small intestine. Localization pattern appears to vary in different species. MRP2 is predominantly localized to the apical membrane of polarized cells.
Multidrug resistance associated protein 2 (MRP2) is expressed in endothelial cells of the blood-brain barrier, various carcinoma cell lines and tumors. The gene encoding MRP2 is localized on human chromosome 10q24 and consists of 32 exons.

Specificity

Additional lower bands including an approx. 175 kDa band representing an immature unglycosylated form may be detected in various extract preparations.

Immunogen

synthetic C-terminal peptide corresponding to amino acids 1528-1545 of human MRP2 conjugated to KLH.

Application

Anti-MRP2 antibody produced in rabbit has been used in:
  • immunoblotting
  • western blotting
  • immunofluorescence

Biochem/physiol Actions

Multidrug Resistance-associated Protein 2 (MRP2) transports endogenous and exogenous anionic conjugates from hepatocytes to the bile. Thus, it contributes to bile flow and plays a role in detoxification and defense against oxidative stress. Patients with the rare autosomal recessive Dubin-Johnson Syndrome develop a mild liver disease caused by MRP2 deficiency. Up regulation of MRP2 expression may be found in hepatocellular carcinomas.
Multidrug resistance associated protein 2 (MRP2) is a transporter which is involved in the efflux of various xenobiotic compounds like doxorubicin and methotrexate.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Schaarschmidt et al.
Theranostics, 8(14), 3766-3780 (2018-08-08)
Rationale: The liver is a central organ not only for metabolism but also immune function. Life-threatening infections of both bacterial and fungal origin can affect liver function but it is yet unknown whether molecular changes differ depending on the pathogen.
Analysis of ATP-binding cassette transporter expression in drug-selected cell lines by a microarray dedicated to multidrug resistance
Annereau JP, et al.
Molecular Pharmacology, 66(6), 1397-1405 (2004)
Biliary elimination of pemetrexed is dependent on Mrp2 in rats: Potential mechanism of variable response in nonalcoholic steatohepatitis
Dzierlenga Al, et al.
Journal of Pharmacology and Experimental Therapeutics, 358(2), 246-253 (2016)
Peter Recknagel et al.
PLoS medicine, 9(11), e1001338-e1001338 (2012-11-16)
Hepatic dysfunction and jaundice are traditionally viewed as late features of sepsis and portend poor outcomes. We hypothesized that changes in liver function occur early in the onset of sepsis, yet pass undetected by standard laboratory tests. In a long-term
Yabin Duan et al.
Drug metabolism and disposition: the biological fate of chemicals, 50(2), 174-186 (2021-12-01)
Hypoxia is the main characteristic of a high-altitude environment, affecting drug metabolism. However, so far, the mechanism of microRNA (miRNA) involved in the regulation of drug metabolism and transporters under high-altitude hypoxia is still unclear. This study aims to investigate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service