Skip to Content
Merck
All Photos(1)

Key Documents

M7383

Sigma-Aldrich

Malic Dehydrogenase from porcine heart

≥400 units/mg protein (biuret), ammonium sulfate suspension

Synonym(s):

L-Malate: NAD+ oxidoreductase, MDH, Malate Dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

ammonium sulfate suspension

specific activity

≥400 units/mg protein (biuret)

foreign activity

Glutamic-Oxalacetic Transaminase ≤0.01%
Glutamic-Pyruvic Transaminase ≤0.01%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Cytoplasmic Isozyme

Application

Malic dehydrogenase has been used in a study to assess the use of a long-wavelength homogeneous enzyme immunoassay for the determination of amikacin in water samples. It has also been used in a study to investigate the lipid metabolism in a liver exposed to cadmium.

Biochem/physiol Actions

The surfactant cetyl (hexadecyl) trimethylammonium bromide inhibits the cytoplasmic isoform of porcine heart malate dehydrogenase but has no effect on the mitochondrial form. SDS however; inhibits both isoforms.

Unit Definition

One unit will convert 1.0 μmole of oxalacetate and β-NADH to L-malate and β-NAD per min at pH 7.5 at 25 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4, 0.1 M KH2PO4, pH 7.0

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M L Sánchez-Martínez et al.
Talanta, 78(1), 305-309 (2009-01-29)
A simple and rapid homogeneous enzyme immunoassay involving the use of the malic dehydrogenase enzyme and a long-wavelength fluorophor, the oxazine Cresyl Violet, is proposed for the determination of the antibiotic amikacin in water samples. An enzymatic tracer has been
P Benoist et al.
Analytical biochemistry, 187(2), 337-344 (1990-06-01)
Nanogram amounts of soluble aminopeptidases, dehydrogenases, and esterases were detected by nondenaturing ultralow gelling point agarose-polyacrylamide gel electrophoresis (ULGA-PAGE). Cytosolic fractions from Frankia sp. were electrophoresed at 4 degrees C in the presence of Co2+, Zn2+, or Mg2+ ions. Then
K Smith et al.
Biochimica et biophysica acta, 884(1), 109-118 (1986-10-29)
The cationic surfactant, cetyl (hexadecyl) trimethylammonium bromide (CTAB), completely inactivates porcine heart cytoplasmic malate dehydrogenase (L-malate:NAD+ oxidoreductase, EC 1.1.1.37) at concentrations (of surfactant) which do not affect the activity of the mitochondrial isoenzyme. These concentrations are close to, or higher
Marie Knockaert et al.
The Journal of biological chemistry, 277(28), 25493-25501 (2002-04-20)
Numerous inhibitors of cyclin-dependent kinases and glycogen synthase kinase-3 (GSK-3) are being developed in view of their potential applications against cancers and neurodegenerative disorders. Among these, paullones constitute a family of potent and apparently selective cyclin-dependent kinase and GSK-3 inhibitors.
Ethel V Larregle et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46(5), 1786-1792 (2008-03-11)
We investigated the effect of exposition to cadmium (Cd, 15ppm for 8 weeks) through drinking water on liver lipid metabolism in adult male Wistar rats. As compared to metal non-exposed (control) rats, the serum triglycerides, cholesterol and LDL+VLDL cholesterol concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service