Recommended Products
Assay
≥98% (TLC)
form
powder
solubility
1 M NaOH: 50 mg/mL, clear, colorless to faintly yellow
storage temp.
−20°C
SMILES string
CN1C(=O)NC=CC1=O
InChI
1S/C5H6N2O2/c1-7-4(8)2-3-6-5(7)9/h2-3H,1H3,(H,6,9)
InChI key
VPLZGVOSFFCKFC-UHFFFAOYSA-N
General description
3-Methyluracil is a methylated uracil or a uracil derivative.
Application
3-Methyluracil (3-MeU) is used along with other methyl-pyrimidines to study relative physical chemical parameters.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Physical chemistry chemical physics : PCCP, 12(19), 4915-4923 (2010-05-07)
In this work we investigated the lowest-lying electronic excitations for a series of methyl-substituted uracil derivatives, i.e., uracil, 1-methyluracil, 3-methyluracil, thymine, 1-methylthymine, 1,3-dimethyluracil, 3-methylthymine, 1,3-dimethylthymine, and their microhydrated complexes by means of coupled cluster singles and approximate doubles (CC2) and
British journal of clinical pharmacology, 55(1), 62-67 (2003-01-22)
(i) To compare the phenotyping of healthy subjects for NAT2 and CYP1A2 activities with caffeine, by the simultaneous assay of the urinary metabolites AFMU and AAMU, and (ii) to ascertain whether NAT2 and CYP1A2 phenotyping is influenced by the use
Radiation research, 114(2), 207-214 (1988-05-01)
Using a pulse radiolysis technique, some nucleic base radicals were produced by the reactions of sulfate radical, SO4-, with 1-, 3-, 5-, and 6-methyluracils, and their optical and kinetic natures were observed. All of their absorption spectra showed main peaks
FEBS letters, 582(23-24), 3313-3319 (2008-09-09)
The human obesity susceptibility gene, FTO, encodes a protein that is homologous to the DNA repair AlkB protein. The AlkB family proteins utilize iron(II), alpha-ketoglutarate (alpha-KG) and dioxygen to perform oxidative repair of alkylated nucleobases in DNA and RNA. We
Journal of the American Chemical Society, 131(51), 18376-18385 (2009-11-26)
The gas-phase substitution reactions of methyl chloride and 1,3-dimethyluracil (at the N1-CH(3)) are examined computationally and experimentally. It is found that, although hydrochloric acid and 3-methyluracil are similar in acidity, the leaving group abilities of chloride and N1-deprotonated 3-methyluracil are
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service