M5795
Anti-MAP2K2/MEK2 Antibody
rabbit polyclonal
Synonym(s):
Anti-MAPKK, Anti-MEK
About This Item
Recommended Products
Product Name
Anti-MAP Kinase Kinase (MEK, MAPKK) antibody produced in rabbit, whole antiserum
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
whole antiserum
antibody product type
primary antibodies
clone
polyclonal
mol wt
antigen 45 kDa (MEK1a)
antigen 46 kDa (MEK2)
contains
15 mM sodium azide
species reactivity
mouse, human, rat
technique(s)
microarray: suitable
western blot: 1:20,000 using rat brain extract and mouse NIH 3T3 fibroblast lysate
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MAP2K1(5604) , MAP2K2(5605)
mouse ... Map2k1(26395) , Map2k2(26396)
rat ... Map2k1(170851) , Map2k2(58960)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Specificity
Immunogen
Application
Biochem/physiol Actions
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service