Skip to Content
Merck
All Photos(1)

Key Documents

M4699

Sigma-Aldrich

MTEP hydrochloride

≥98% (HPLC)

Synonym(s):

MTEP hydrochloride, 3-[(2-methyl-1,3-thiazol-4-yl)ethynyl]pyridine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8N2SHCl
CAS Number:
Molecular Weight:
236.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

storage condition

desiccated

color

white to beige

solubility

H2O: 30 mg/mL, clear

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

2-8°C

SMILES string

Cl.Cc1nc(cs1)C#Cc2cccnc2

InChI

1S/C11H8N2S.ClH/c1-9-13-11(8-14-9)5-4-10-3-2-6-12-7-10;/h2-3,6-8H,1H3;1H

InChI key

YCIOJDKGCWAHLR-UHFFFAOYSA-N

Biochem/physiol Actions

MTEP is a potent and highly selective antagonist for mGluR5.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Frankowska et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 71(4) (2020-12-15)
The abundance of research indicates that enriched environment acts as a beneficial factor reducing the risks of relapse in substance use disorder. There is also strong evidence showing the engagement of brain dopaminergic and glutamatergic signaling through the dopamine D2-like
Rianne R Campbell et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(14), 2745-2761 (2019-02-10)
The bed nucleus of the stria terminalis (BNST) is part of the limbic-hypothalamic system important for behavioral responses to stress, and glutamate transmission within this region has been implicated in the neurobiology of alcoholism. Herein, we used a combination of
Tomas Ondrejcak et al.
Neurobiology of disease, 127, 582-590 (2019-03-27)
Soluble synaptotoxic aggregates of the main pathological proteins of Alzheimer's disease, amyloid β-protein (Aß) and tau, have rapid and potent inhibitory effects on long-term potentiation (LTP). Although the promotion of synaptic weakening mechanisms, including long-term depression (LTD), is posited to
Richard M Cleva et al.
Frontiers in pharmacology, 2, 93-93 (2012-01-11)
Pharmacological manipulation of the type 5 metabotropic glutamate (mGlu5) receptor alters various addiction related behaviors such as drug self-administration and the extinction and reinstatement of drug-seeking behavior. However, the effects of pharmacological modulation of mGlu5 receptors on brain reward function
Jongyun Myeong et al.
Cell reports, 41(11), 111820-111820 (2022-12-15)
Synaptic facilitation is a major form of short-term plasticity typically driven by an increase in residual presynaptic calcium. Using near-total internal reflection fluorescence (near-TIRF) imaging of single vesicle release in cultured hippocampal synapses, we demonstrate a distinctive, release-dependent form of

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service