Skip to Content
Merck
All Photos(1)

Key Documents

M4530

Sigma-Aldrich

Medium 199

Medium 199
1 of 1 reviewers received a sample product or took part in a promotion

With Earle′s salts, ʟ-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

M199 Medium, TCM 199

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

Product Name

Medium 199, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

NaHCO3: yes
L-glutamine: yes
HEPES: no
Earle’s salts (5% CO2): yes
phenol red: yes
sodium pyruvate: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Complete liquid version of the original M 199 formulation published by Morgan.
Medium 199 is a combination of vitamins, amino acids, and other factors that were originally developed as a completely defined media formulation for the culture of primary explants. This medium, when properly supplemented has broad applicability, particularly for non-transformed cells. Medium 199 is widely used for vaccine production, in vitro cultivation of primary pancreatic explants, and lens tissues.

Application

Medium 199 has been used:
  • as a suspension media to maintain the primary neuronal cultures
  • as a component of the collagen gel for ex vivo embryonic heart culture
  • to maintain the ovarian tissue sections as part of the slow-freezing protocol

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Ma et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 72(4), 489-498 (2016-08-28)
We previously reported that pretreatment with the potent antioxidant TEMPOL improves mitochondrial function and restores preconditioning in the aging heart. Because mitophagy is implicated in cardiac preconditioning and declines with age, this study was designed to investigate how age influences
Patrick C Baer et al.
International journal of molecular sciences, 19(6) (2018-06-13)
Stem cell-based therapies require cells with a maximum regenerative capacity in order to support regeneration after tissue injury and organ failure. Optimization of this regenerative potential of mesenchymal stromal/stem cells (MSC) or their conditioned medium by in vitro preconditioning regimens
Vladimir Jovasevic et al.
Journal of virology, 82(7), 3311-3319 (2008-01-25)
In this report we propose a model in which after the herpes simplex virus (HSV) capsid docks at the nuclear pore, the tegument protein attached to the capsid must be cleaved by a serine or a cysteine protease in order
Amanda Fanous et al.
Plastic and reconstructive surgery, 143(3), 760-767 (2019-01-03)
Keloid scarring is a serious condition that mostly affects patients of African or Asian descent. Often disfiguring, this condition can have devastating psychosocial consequences. To date, no treatment modality has been proven ideal. The authors' objectives were (1) to determine
Immacolata Castellano et al.
Oxidative medicine and cellular longevity, 2018, 2087373-2087373 (2018-06-01)
Chronic hyperglycemia is associated with oxidative stress and vascular inflammation, both leading to endothelial dysfunction and cardiovascular disease that can be weakened by antioxidant/anti-inflammatory molecules in both healthy and diabetic subjects. Among natural molecules, ovothiol A, produced in sea urchin

Articles

Medium 199 supports non-transformed cell cultivation in virology and vaccine production, offering broad species applicability.

Medium 199 supports non-transformed cell cultivation in virology and vaccine production, offering broad species applicability.

Medium 199 supports non-transformed cell cultivation in virology and vaccine production, offering broad species applicability.

Medium 199 supports non-transformed cell cultivation in virology and vaccine production, offering broad species applicability.

See All

Related Content

Discover the formulation for Medium 199 for cell culture applications, including vaccine development.

Discover the formulation for Medium 199 for cell culture applications, including vaccine development.

Discover the formulation for Medium 199 for cell culture applications, including vaccine development.

Discover the formulation for Medium 199 for cell culture applications, including vaccine development.

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. London, U.K.
    • Reviews 3
    • Votes 4
    5 out of 5 stars.

    Good value for money and can be used as the basal medium for endothelial cell growth

    Really good value for money and always arrives with a good shelf-life in terms of expiry date. Great for adding cytokines + hydrocortisone to to create your own endothelial cell growth medium - has kept my HUVECs consistently happy since I started using it in 2018. Also great to use when isolating human primary endothelial cells.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service