Skip to Content
Merck
All Photos(1)

Key Documents

M4074

Sigma-Aldrich

Mometasone furoate

≥98% (HPLC)

Synonym(s):

(11β,16α)-9,21-Dichloro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30Cl2O6
CAS Number:
Molecular Weight:
521.43
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +50 to +60°, c = 0.5 in methanol

color

white to off-white

solubility

DMSO: ≥20 mg/mL

originator

Schering Plough

storage temp.

2-8°C

SMILES string

C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5ccco5)C(=O)CCl

InChI

1S/C27H30Cl2O6/c1-15-11-19-18-7-6-16-12-17(30)8-9-24(16,2)26(18,29)21(31)13-25(19,3)27(15,22(32)14-28)35-23(33)20-5-4-10-34-20/h4-5,8-10,12,15,18-19,21,31H,6-7,11,13-14H2,1-3H3/t15-,18+,19+,21+,24+,25+,26+,27+/m1/s1

InChI key

WOFMFGQZHJDGCX-ZULDAHANSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Application

Mometasone furoate has been used to study its effect on Leishmania major amastigotes.

Biochem/physiol Actions

Mometasone furoate is a 17-heterocyclic intranasal corticosteroid. It is effectively used as a first-line daily intranasal therapeutic for allergic rhinitis and nasal polyposis. Mometasone furoate is also used as an adjunct to anti-bacterials for treating acute rhinosinusitis. In addition, it relieves the symptoms of asthma in both adults and adolescents by exhibiting a broad spectrum of anti-inflammatory properties.
Mometasone furoate is an anti-inflammatory glucocorticoid.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Schering Plough. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David W Kennedy
Expert review of respiratory medicine, 6(5), 493-498 (2012-11-09)
Chronic rhinosinusitis is widely recognized as one of the most common chronic disease entities. Since its introduction in the USA in 1985, the role of functional endoscopic sinus surgery as an adjunct to medical therapy in the treatment of chronic
An In Vitro Investigation of the Effects of the Glucocorticoid on Leishmania major Amastigotes
Renani EH, et al.
Avicenna journal of phytomedicine, 6(4), 127-132 (2019)
Inhaled Mometasone Furoate
Sharpe M and Jarvis B
Drugs, 61(9), 1325-1350 (2001)
Mohamed A Bitar et al.
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery, 270(3), 931-937 (2012-09-27)
This study aimed at observing the efficacy of mometasone fuorate monohydrate nasal spray on obstructive adenoids in children and identifying the characteristics of responders using a pilot study including children aged 2-11 years, with evidence of more than 50 %
Johanna Svensson et al.
Primary care respiratory journal : journal of the General Practice Airways Group, 21(4), 412-418 (2012-09-25)
Acute rhinosinusitis is a common disease with an increasing incidence rate. It causes substantial costs to the individual and to society through healthcare consumption and absence from work. The use of antibiotics is widespread in the treatment of acute rhinosinusitis

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service