Skip to Content
Merck
All Photos(1)

Key Documents

M3781

Sigma-Aldrich

6α-Methylprednisolone 21-hemisuccinate sodium salt

lyophilized powder

Synonym(s):

6α-Methylprednisolone sodium succinate, MPSL, 6α-Methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 21-hemisuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C26H33O8Na
CAS Number:
Molecular Weight:
496.53
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

sterility

non-sterile

Assay

≥98.00% (TLC)

form

lyophilized powder

technique(s)

toxicology assay: suitable

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].C[C@H]1CC2C3CC[C@](O)(C(=O)COC(=O)CCC([O-])=O)[C@@]3(C)C[C@H](O)C2[C@@]4(C)C=CC(=O)C=C14

InChI

1S/C26H34O8.Na/c1-14-10-16-17-7-9-26(33,20(29)13-34-22(32)5-4-21(30)31)25(17,3)12-19(28)23(16)24(2)8-6-15(27)11-18(14)24;/h6,8,11,14,16-17,19,23,28,33H,4-5,7,9-10,12-13H2,1-3H3,(H,30,31);/q;+1/p-1/t14-,16-,17-,19-,23+,24-,25-,26-;/m0./s1

InChI key

FQISKWAFAHGMGT-SGJOWKDISA-M

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Application

6α-methylprednisolone 21-hemisuccinate sodium salt has been used to investigate glucocorticoid-induced osteonecrosis under hypoxic conditions in the MC3T3-E1 osteoblast cell line using a cell cytotoxicity assay, flow cytometry and western blotting.

Biochem/physiol Actions

Glucocorticoid with slightly longer half-life than that of prednisolone.

Physical form

Lyophilized containing ~5% phosphate buffer salts.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[The issue of methylprednisolone sodium succinate dosage for use].
Yu-cai Zhang et al.
Zhonghua er ke za zhi = Chinese journal of pediatrics, 49(5), 389-390 (2011-06-01)
Julie M Jones et al.
The Journal of the American Osteopathic Association, 112(2), 90-92 (2012-02-15)
Hypocomplementemic urticarial vasculitis syndrome (HUVS) is a type III hypersensitivity reaction characterized by urticaria with persistent acquired hypocomplementemia. Although HUVS is uncommon, it is important for physicians to be familiar with this disease, as the initial presentation is often life-threatening
Dapsone hypersensitivity syndrome complicated by Scedosporium apiospermum pneumonia in an immunocompetent patient.
L Ceccarelli et al.
Infection, 40(4), 459-462 (2011-11-30)
Rina Ulmansky et al.
Journal of controlled release : official journal of the Controlled Release Society, 160(2), 299-305 (2012-01-10)
We have previously shown that intravenous (i.v.) treatment with sterically stabilized nano-liposomes (NSSL) actively remote-loaded with the glucocorticoid (GC) methylprednisolone hemisuccinate (NSSL-MPS) or betamethasone hemisuccinate (NSSL-BMS) significantly decreased severity of adjuvant arthritis in Lewis rats (a model of human rheumatoid
Takaaki Kitakawa et al.
Documenta ophthalmologica. Advances in ophthalmology, 124(3), 249-254 (2012-03-10)
The purpose of this study was to report a patient with acute zonal occult outer retinopathy (AZOOR) unilaterally, who received steroid pulse therapy. A 42-year-old woman presented with photopsias and severe vision loss in her left eye. Visual acuity was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service