Skip to Content
Merck
All Photos(1)

Key Documents

M2824

Sigma-Aldrich

Meptazinol hydrochloride

Synonym(s):

3-(3-ethylhexahydro-1-methyl-1H-azepin-3-yl)-phenol hydrochloride, IL-22811 hydrochloride, WY-22811 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO· HCl
CAS Number:
Molecular Weight:
269.81
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

color

white to off-white

solubility

H2O: >10 mg/mL

originator

Wyeth

SMILES string

Cl.CCC1(CCCCN(C)C1)c2cccc(O)c2

InChI

1S/C15H23NO.ClH/c1-3-15(9-4-5-10-16(2)12-15)13-7-6-8-14(17)11-13;/h6-8,11,17H,3-5,9-10,12H2,1-2H3;1H

InChI key

MPJUSISYVXABBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Meptazinol entered the human pharmaceutical market as a racemic mixture in the 1980s for use as an analgesic. Its pharmacology is not completely understood; however, its analgesic properties are mostly due to its partial agonism at the mu1 opioid receptor. Due to its partial agonism, Meptazinol antagonizes morphine dependence in vivo. Its advantage over other opiates is its reduced capacity to cause addition and respiratory depression, also due to its intrinsic activity as a partial agonist. Meptazinol has been found to have additional activity as an acetylcholinesterase (AChE) inhibitor, particularly in its (-) enantiomeric form, which may partially explain its analgesic properties. AChE inhibitors are used to treat Alzheimer′s disease, providing additional interest in this compound.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin-xing Ge et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 881-882, 126-130 (2011-12-30)
A rapid, simple and sensitive LC-MS/MS method was developed and validated for the determination of Bis(9)-(-)-Meptazinol (B9M) in rat plasma. Protein precipitation method was used for sample preparation, using five volumes of methanol as the precipitation agent. The analytes were
Meiyan Lu et al.
Bioorganic & medicinal chemistry letters, 15(10), 2607-2609 (2005-05-03)
Three meptazinol benzoyl esters (1-3) were synthesized as prodrugs to minimize the first-pass effect of meptazinol and improve the bioavailability. Among these three esters, compound 3 showed better bioavailability than the parent meptazinol. Further, the relative regional bioavailability of prodrug
Wei Li et al.
Acta pharmacologica Sinica, 26(3), 334-338 (2005-02-18)
To investigate the mechanism of action of a potent analgesic, (+/-)-meptazinol. The structures of meptazinol enantiomers were compared with opioid pharmacophore and tramadol. Neither enantiomer of meptazinol fitted any patterns among the opioid pharmacophore and tramadol, although they did share
J B Madsen et al.
Acta anaesthesiologica Scandinavica, 37(3), 307-313 (1993-04-01)
A technique of epidural catheterization in rabbits is described. Twelve albino rabbits received a totally implanted epidural catheter system. The system was implanted surgically, and the functioning of the system tested for a period of 3 months. X-ray examinations following
Aviv Paz et al.
Journal of medicinal chemistry, 52(8), 2543-2549 (2009-03-31)
A bis-(-)-nor-meptazinol derivative in which the two meptazinol rings are linked by a nonamethylene spacer is a novel acetylcholinesterase inhibitor that inhibits both catalytic activity and Abeta peptide aggregation. The crystal structure of its complex with Torpedo californica acetylcholinesterase was

Articles

We offers many products related to opioid receptors for your research needs.

We offers many products related to opioid receptors for your research needs.

We offers many products related to opioid receptors for your research needs.

We offers many products related to opioid receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service