Skip to Content
Merck
All Photos(3)

Key Documents

M2574

Sigma-Aldrich

Meropenem trihydrate

≥98% (HPLC), powder, antibacterial agent

Synonym(s):

(1R,5S,6S)-2-[(3S,5S)-5-(dimethylaminocarbonyl)pyrrolidin-3-ylthio]-6-[(R)-1-hydroxyethyl]-1-methylcarbapen-2-em-3-carboxylic acid trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25N3O5S · 3H2O
CAS Number:
Molecular Weight:
437.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Meropenem trihydrate, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: ≥20 mg/mL

originator

AstraZeneca

storage temp.

−20°C

SMILES string

O.O.O.C[C@@H](O)[C@@H]1[C@H]2[C@@H](C)C(S[C@@H]3CN[C@@H](C3)C(=O)N(C)C)=C(N2C1=O)C(O)=O

InChI

1S/C17H25N3O5S.3H2O/c1-7-12-11(8(2)21)16(23)20(12)13(17(24)25)14(7)26-9-5-10(18-6-9)15(22)19(3)4;;;/h7-12,18,21H,5-6H2,1-4H3,(H,24,25);3*1H2/t7-,8-,9+,10+,11-,12-;;;/m1.../s1

InChI key

CTUAQTBUVLKNDJ-OBZXMJSBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Meropenem trihydrate (MRP) belongs to the carbapenem group of compounds. It is susceptible to degradation at high temperature and humidity.[1] It is soluble in methanol-water or acetone-water combinations. It is used as an antibacterial agent for treating meningitis and pneumonia.[2]

Application

Meropenem trihydrate has been used in the
  • antibiotic susceptibility testing of E coli isolates from harbor estuary sediment samples[3]
  • in the bacterial killing assay and for screening antibiotic resistance of cystic fibrosis patient′s sputum based Staphylococcus aureus transformed macrophages in phagocyte infection[4]
  • for screening Enterococcus faecalis from human bile[5]

Biochem/physiol Actions

Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.
Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic.

Features and Benefits

This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa Kolano et al.
Chembiochem : a European journal of chemical biology, 21(18), 2628-2634 (2020-04-16)
Proline-rich antimicrobial peptides expressed in insects are primarily active against Enterobacteriaceae. Mechanistically, they target the bacterial (70S) ribosome after partially transporter-based cellular uptake, as revealed for Api137 and Onc112 on Escherichia coli. Following molecular modeling indicating that the Onc112 contact
Thanh C Tran et al.
MicrobiologyOpen, 9(12), e1130-e1130 (2020-11-10)
To assess a cost-effective in-house selective plate formula for actively screening carbapenem-resistant Enterobacteriaceae (CRE). The in-house formula included CHROMagarTM Orientation, meropenem, and ingredients present in the Mac-Conkey formula, such as bile salts and crystal violet (pH 6.9-7.2). American Type Culture
Measurement and correlation of solubility of meropenem trihydrate in binary (water+ acetone/tetrahydrofuran) solvent mixtures
Zhou L, et al.
Chinese Journal of Chemical Engineering, 25(10), 1461-1466 (2017)
Human bile reduces antimicrobial activity of selected antibiotics against Enterococcus faecalis and Escherichia coli in vitro
Wulkersdorfer B, et al.
Antimicrobial agents and chemotherapy, 61(8), e00527-e00517 (2017)
The Radiostability of Meropenem Trihydrate in Solid State
Kilinska K, et al.
Molecules (Basel), 23(11), 2738-2738 (2018)

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Questions

  1. Hello, What is the potency of Meropenem trihydrate (M2574)

    1 answer
    1. The potency of this product has not been determined. The minimum purity of this material is 97.5% by HPLC. Please see the link below to review a sample or lot specific Certificate of Analysis:
      https://www.sigmaaldrich.com/product/sigma/m2574#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service