Skip to Content
Merck

M2147

Mevinolin

≥98% (HPLC), HMG-CoA reductase inhibitor, crystalline powder

Synonym(s):

2-Methyl-1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-(tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl)ethyl]-1-naphthalenyl ester butanoic acid, 6-α-Methylcompactin, Lovastatin, Monacolin K

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H36O5
CAS Number:
Molecular Weight:
404.54
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Mevinolin from Aspergillus sp., ≥98% (HPLC)

Quality Level

assay

≥98% (HPLC)

form

crystalline powder

mp

175.4 °C

solubility

ethanol: 9.80-10.20 mg/mL, clear, colorless to faintly yellow

originator

Abbott

storage temp.

2-8°C

SMILES string

[H][C@]12[C@H](C[C@@H](C)C=C1C=C[C@H](C)[C@@H]2CC[C@@H]3C[C@@H](O)CC(=O)O3)OC(=O)[C@@H](C)CC

InChI

1S/C24H36O5/c1-5-15(3)24(27)29-21-11-14(2)10-17-7-6-16(4)20(23(17)21)9-8-19-12-18(25)13-22(26)28-19/h6-7,10,14-16,18-21,23,25H,5,8-9,11-13H2,1-4H3/t14-,15-,16-,18+,19+,20-,21-,23-/m0/s1

InChI key

PCZOHLXUXFIOCF-BXMDZJJMSA-N

Gene Information

human ... HMGCR(3156)
rat ... Hmgcr(25675)

Biochem/physiol Actions

Lovastatin is a cholesterol lowering drug and competitive inhibitor of HMG-CoA reductase, a rate limiting enzyme in cholesterol synthesis. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. This product is a substrate of Pgp and CYP3A. It increases cellular resistance to anticancer agents such as doxorubicin and induces apoptosis in myeloma cells. The roles of Pgp and CYP3A, possible connection between drug resistance, regulation of the mevalonate pathway, and iosprenylation of signaling proteins in these observations remain to be resolved. The product induces apoptosis in numerous cancer cell lines perhaps, in part, by inhibiting the isoprenylation of Rho family GTPases. It causes cell cycle arrest in G1 and G2/M phases.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


pictograms

Health hazard

signalword

Warning

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Wen-Bin Zhong et al.
Endocrinology, 144(9), 3852-3859 (2003-08-23)
Lovastatin has been used to treat hypercholesterolemia through blocking the mevalonate biosynthesis pathway. Inhibition of mevalonate synthesis may result in antiproliferation and cell apoptosis. The aim of the present study was to examine the apoptotic effect of lovastatin in human
N W C J van de Donk et al.
Leukemia, 16(7), 1362-1371 (2002-07-03)
Lovastatin is an irreversible inhibitor of HMG-CoA reductase and blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. Isoprenylation of target proteins, like the GTP-binding protein Ras, is essential for their membrane localization and
Kimberly A Hartwell et al.
Nature chemical biology, 9(12), 840-848 (2013-10-29)
Efforts to develop more effective therapies for acute leukemia may benefit from high-throughput screening systems that reflect the complex physiology of the disease, including leukemia stem cells (LSCs) and supportive interactions with the bone marrow microenvironment. The therapeutic targeting of



Global Trade Item Number

SKUGTIN
M2147-25MG04061834041641