Skip to Content
Merck
All Photos(1)

Key Documents

M104

Sigma-Aldrich

(±)-Muscarine chloride hydrate

≥98% (HPLC)

Synonym(s):

Tetrahydro-4-hydroxy-N,N,N,5-tetramethyl-2-furanmethanammonium chloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20ClNO2 · xH2O
CAS Number:
Molecular Weight:
209.71 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

deionized water: ≥20 mg/mL

storage temp.

room temp

SMILES string

[Cl-].CC1OC(CC1O)C[N+](C)(C)C

InChI

1S/C9H20NO2.ClH/c1-7-9(11)5-8(12-7)6-10(2,3)4;/h7-9,11H,5-6H2,1-4H3;1H/q+1;/p-1

InChI key

WUFRNEJYZWHXLC-UHFFFAOYSA-M

Application

(±)-Muscarine chloride hydrate may be used:
  • as an acetylcholine receptor agonist in neuronal precursor cell lines PC 12 cells
  • as toxin test compound for paper spray mass spectrometry (PSMS) optimization experiments
  • as a component of quality control(QC) in hydrophilic interaction liquid chromatography-high resolution tandem mass spectrometry (HILIC-HRMS/MS) analysis of human urine samples

Biochem/physiol Actions

Muscarine is a potent agonist than acetylcholine and is not susceptible to degradation by cholinesterases. Muscarine administration results in muscle spasm especially in gut, bronchus and uterus.
Muscarinic acetylcholine receptor agonist; originally isolated from Amanita muscaria.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thomas P Bambauer et al.
Talanta, 204, 677-684 (2019-07-31)
Paper spray mass spectrometry (PSMS) is currently used in different analytical fields, but less effort has been made so far to use PSMS for highly polar compounds. Such analytes usually show poor performance in PSMS due to their high affinity
P J FRASER
British journal of pharmacology and chemotherapy, 12(1), 47-52 (1957-03-01)
The action of chromatographically pure crystalline muscarine chloride, prepared from Amanita muscaria, has been compared with acetylcholine chloride (ACh) on a number of different organs from a variety of species. Muscarine caused spasm in vivo and in vitro of muscles
Chapter 67 - Mushroom Toxins
Puschner B
Veterinary Toxicology, 955-966 (2018)
Two-dimensional surface plasmon resonance imaging system for cellular analysis
Biosensors and Biodetection, 12(1), 47-52 (2017)
Thomas P Bambauer et al.
Talanta, 213, 120847-120847 (2020-03-24)
The analytical proof of a toxic mushroom and/or plant ingestion at an early stage of a suspected intoxication can be crucial for fast therapeutic decision making. Therefore, comprehensive analytical procedures need to be available. This study aimed to develop a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service