Skip to Content
Merck
All Photos(1)

Key Documents

M0800

Sigma-Aldrich

Anti-p38 MAP Kinase antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-ERK, Anti-ERK-2, Anti-ERK2, Anti-ERT1, Anti-MAPK2, Anti-NS13, Anti-P42MAPK, Anti-PRKM1, Anti-PRKM2, Anti-p38, Anti-p40, Anti-p41, Anti-p41mapk, Anti-p42-MAPK

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 40 kDa

species reactivity

mouse, rat

technique(s)

microarray: suitable
western blot: 1:10,000 using NIH3T3 mouse fibroblast lysate and rat brain extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPK1(5594)
mouse ... Mapk1(26413)
rat ... Mapk1(116590)

General description

P38 MAPK is a mitogen-activated protein kinase (MAPK) that belongs to serine/threonine protein kinases family and is activated in response to stress signals like- osmolarity changes, UV-irradiation, heat shock,cytokines and lipopolysaccharides (LPS). It mainly expresses in brain, muscle, thymus, spleen, heart, lung, kidney and fibroblasts.

Specificity

Rabbit anti-p38 MAP Kinase antibody reacts specifically with p38 MAP kinase from mouse and rat.

Immunogen

synthetic peptide K-DEVISFVPPPLDQEEMES derived from the C-terminus of mouse p38 MAP kinase (amino acids 343-360 with N-terminally added lysine) as immunogen. This sequence is highly conserved in human and rat p38 MAP kinase.

Application

Anti-p38 MAP Kinase antibody has been used in western blotting (diluted 1:1000) using using NIH 3T3 mouse fibroblasts cell lysate and rat brain extract. It has also been used in microarray.

Biochem/physiol Actions

Mitogen-activated protein kinases (MAP kinases, MAPKs) are a family of protein kinases which are considered to play a crucial role in signal transduction pathways in mammalian cells by leading mitogenic signals to their intracellular targets. MAP kinases regulate several cellular processes, such as proliferation, differentiation, response to stress and oncogenesis. p38 MAPK is activated in response to stress signals like- osmolarity changes, ultravoilet-irradiation, heat shock, cytokines and lipopolysaccharides (LPS). It is required for phosphorylation of MAPK-activated protein kinase 2 (MAPKAPK-2). Anti-p38 MAP Kinase antibody can be used to study the differential tissue expression, specific activation requirements and intracellular localization of MAP kinase isoforms in normal and neoplastic tissues.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jarod Li et al.
The Journal of biological chemistry, 289(51), 35633-35643 (2014-11-06)
A growing body of work has shown that the highly homologous T-box transcription factors TBX2 and TBX3 play critical but distinct roles in embryonic development and cancer progression. For example, TBX2 and TBX3 are up-regulated in several cancers and recent
Brittany-Amber Jacobs et al.
Scientific reports, 8(1), 11547-11547 (2018-08-03)
Persistent infection with human papillomavirus (HPV) is responsible for nearly all new cervical cancer cases worldwide. In low- and middle-income countries (LMIC), infection with helminths has been linked to increased HPV prevalence. As the incidence of cervical cancer rises in
Jarod Li et al.
Molecular biology of the cell, 24(22), 3569-3576 (2013-09-13)
The T-box transcription factor, TBX3, plays an important role in embryonic development, and haploinsufficiency of TBX3 causes ulnar-mammary syndrome. Overexpression of TBX3, on the other hand, is associated with several cancers, and preliminary evidence suggests that increased levels of TBX3
Soumyajit Banerjee Mustafi et al.
Cell stress & chaperones, 14(6), 579-589 (2009-03-18)
Chinese hamster lung fibroblasts V79 cells were treated with heat stress for 4 weeks with short duration (15 min) heat shock every alternate day in culture. It was observed that Hsp 70 and the antioxidant enzyme MnSOD became overexpressed during
R Y Liu et al.
The Journal of biological chemistry, 275(28), 21086-21093 (2000-04-28)
To elucidate mechanisms of tumor necrosis factor alpha (TNF-alpha)-induced proliferation of a number of human leukemia and lymphoma cell lines, we examined the role of p38 mitogen-activated protein kinase (MAPK) in TNF-alpha signaling in Mo7e and Hut-78 cells. TNF-alpha-dependent p38

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service