Skip to Content
Merck

LB8

Latex beads, polystyrene

0.8 μm mean particle size

Synonym(s):

Latex Beads, Latex Microspheres, laboratory latex beads, latex sphers

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous suspension

composition

Solids, 10%

packaging

pack of 1 mL, pack of 15 mL, pack of 2 mL

mean particle size

0.8 μm

General description

Polystyrene microparticles are stabilizedcolloidal particles that possess a negative charge and are synthesized by thepolymerization of styrene under conditions that induce spontaneous coalescentbead formation. Due to their versatility, these beads can be used in antibody-mediatedagglutination diagnostics, electron microscopy, cell counter calibration, andphagocytosis experiments.

Application

Polystyrene latex beads have been used:
  • inthe flow cytometric assay of endothelial microparticles (EMPs)
  • in thedetection of capsular polysaccharide (CPS) antibody/antigen latex agglutinationtest
  • to evaluate the phagocytic activity in hemocytes of shrimps


Still not finding the right product?

Explore all of our products under Latex beads, polystyrene


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Humoral immune responses following experimental infection of goats with Mycoplasma capricolum subsp. capri pneumoniae
John B M, et al.
Veterinary Microbiology, 84(1-2) (2002)
Effects of traditional medical herbs ?minor bupleurum decoction? on the non-specific immune responses of white shrimp (Litopenaeus vannamei)
Yu-Sheng, et al.
Fish & Shellfish Immunology, 218-225, 84(1-842) (2017)
AGE/RAGE-Induced EMP Release via the NOX-Derived ROS Pathway
Ying-Hua C, et al.
Journal of Diabetes Research (2018)



Global Trade Item Number

SKUGTIN
LB8-1ML04061838108258
LB8-15ML04061838088147
LB8-100ML04061838208507
LB8-2ML04061838208514