Skip to Content
Merck

LB11

Latex beads, polystyrene

1.1 μm mean particle size

Synonym(s):

Latex Microspheres, Polystyrene Beads, Polystyrene Microspheres, plastic beads

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous suspension

composition

Solids, 10%

packaging

pack of 1 mL, pack of 15 mL, pack of 2 mL

mean particle size

1.1 μm

General description

Latex bead suspensions mostly contain polymer particles and water. It includes small amounts of surfactant, sodium bicarbonate, and potassium sulfate. Polystyrene microparticles are stabilized colloidal particles bearing a negative charge. Polymerization of styrene under certain conditions results in spontaneous coalescent bead formation leading to the formation of microparticles.

Application

Latex beads, polystyrene have been used:
  • in phagocytosis experiments
  • to provide a stimulated condition for adjusting the neutrophil count in cell suspension
  • to incubate macrophages for phagosome trafficking assays

Biochem/physiol Actions

Latex beads (polystyrene) are used in electron microscopy, cell counter calibration, antibody-mediated and agglutination diagnostics.


Still not finding the right product?

Explore all of our products under Latex beads, polystyrene


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Irene Martínez De Toda et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 75(10), 1827-1833 (2019-10-20)
Several parameters of immune function, oxidative, and inflammatory stresses have been proposed as markers of health and predictors of longevity and mortality. However, it is unknown if any of these parameters can be used as predictors of survival in centenarians.
Christin Neuber et al.
Clinical hemorheology and microcirculation, 61(2), 291-299 (2015-09-28)
Survivors of Hodgkin's disease as well as of breast and lung cancer are at risk of radiation-associated cardiovascular disease. Recent studies demonstrated a correlation between cardiovascular risk factors and circulating endothelial microparticles (EMP) and thereby suggest increased EMP levels in
Sabna Rajeev Krishnan et al.
Neoplasia (New York, N.Y.), 18(1), 25-32 (2016-01-26)
The confinement of multiple myeloma (MM) to the bone marrow microenvironment requires an invasive bone marrow biopsy to monitor the malignant compartment. The existing clinical tools used to determine treatment response and tumor relapse are limited in sensitivity mainly because



Global Trade Item Number

SKUGTIN
LB11-15ML04061838208422
LB11-1ML04061838108210
LB11-2ML04061838208439