Skip to Content
Merck

L8286

Ultima Gold LSC Cocktail

For aqueous and non-aqueous samples

Synonym(s):

Scintillation Cocktail

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

composition

bis(2-ethylhexyl)hydrogen phosphate, 10-30 % (w/w) , Bis(isopropyl)naphthalene, 65-85 % (w/w) , Dioctyl sodium sulfosuccinate, 1-5 % (w/w) , Ethoxylated nonylphenol, 10-30 % (w/w) , Triethylphosphate, 1-5 % (w/w)

color

colorless

pH

6.3 (20 °C)

bp

290 °C/1,013 hPa

mp

-30 °C

density

0.960 g/cm3

General description

Ultima Gold LSC Cocktail is a safer line of liquid scintillation cocktails (LSC) designed for a wide range of aqueous solutions. Liquid scintillation counting in radionuclide metrology involves preparing samples which are emulsions of aqueous radioactive solutions in liquid scintillation cocktails. Ultima Gold LSC Cocktail technology is an alternative screening tool to alpha spectrometry and gas flow proportional counting.

Application

Ultima Gold LSC Cocktail is widely used to measure low-energy radionuclides. Metrology laboratories also use it for the standardization of radioactive solutions.

Features and Benefits

Ultima Gold LSC Cocktail has the following benefits:
  • A high flash point (150 °C) offers greater safety.
  • High sample load capacity and high counting efficiency − up to 50% for 3H. 
  • Compatible with alkaline samples.
  • It will not diffuse through polyethylene vials. 
  • Biodegradable. 
  • Compatible with polar organic solvents.

Legal Information

Ultima Gold is a trademark of Revvity


signalword

Danger

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

305.6 °F - closed cup

flash_point_c

152 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter

Hazard Classifications

Aquatic Chronic 1 - Asp. Tox. 1 - ED ENV 1 - Eye Dam. 1 - Skin Corr. 1B



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library