Skip to Content
Merck
All Photos(1)

Key Documents

L7773

Sigma-Aldrich

Lysozyme from chicken egg white

aseptically filled

Synonym(s):

Mucopeptide N-acetylmuramoylhydrolase, Muramidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

sterility

aseptically filled

Quality Level

form

powder

enzyme activity

≥40000 units/mg protein

mol wt

single-chain 14.3 kDa

UniProt accession no.

storage temp.

−20°C

Gene Information

chicken ... LYZ(396218)

Looking for similar products? Visit Product Comparison Guide

Application

Enzyme breaks down the cell walls of bacteria; used to prepare spheroplasts.

Biochem/physiol Actions

Lysozyme hydrolyzes β(1→4) linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin. Gram-positive cells are quite susceptible to this hydrolysis as their cell walls have a high proportion of peptidoglycan. Gram-negative bacteria are less susceptible due to the presence of an outer membrane and a lower proportion of peptidoglycan. However, these cells may be hydrolyzed in the presence of EDTA that chelates metal ions in the outer bacterial membrane.

The enzyme is active over a broad pH range (6.0 to 9.0). At pH 6.2, maximal activity is observed over a wider range of ionic strengths (0.02 to 0.100 M) than at pH 9.2 (0.01 to 0.06 M).

Unit Definition

One unit will lyse 0.6 μg of Micrococcus lysodeikticus per minute by turbidimetric detection at 600 nm when suspended in buffer at pH 6.2 at 25 °C.

Preparation Note

Prepared from L6876

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P F Zabrodskiĭ et al.
Eksperimental'naia i klinicheskaia farmakologiia, 75(11), 19-21 (2013-01-18)
It is established in experiments on noninbred rats that the use of imunofan (20 mg/kg daily) and polyoxidonium (150 mg/kg daily) for 7 days on the background of chronic intoxication with organophosphorus agent VX (0.01 LD50, single daily treatment for
Mariela G Del Giudice et al.
Infection and immunity, 81(3), 956-964 (2013-01-16)
Secretion of proteins in Gram-negative bacteria is a high-energy-consuming process that requires translocation across two membranes and a periplasmic space composed of a mesh-like layer, the peptidoglycan. To achieve this, bacteria have evolved complex secretion systems that cross these barriers
Anne Derbise et al.
The Journal of infectious diseases, 207(10), 1535-1543 (2013-02-14)
Yersinia pestis (the plague bacillus) and its ancestor, Yersinia pseudotuberculosis (which causes self-limited bowel disease), encode putative homologues of the periplasmic lysozyme inhibitor Ivy and the membrane-bound lysozyme inhibitor MliC. The involvement of both inhibitors in virulence remains subject to
Mahdieh Mansouri et al.
Molecules (Basel, Switzerland), 18(1), 789-813 (2013-01-25)
The interactions between cyclophosphamide (CYC) and lysozyme (LYZ) in the presence of different cyclodextrins (CDs) were investigated by UV absorption, fluorescence spectroscopy, circular dichroism (CD), and molecular modeling techniques under imitated physiological conditions. The UV absorption results showed the formation
Zhe Wang et al.
The journal of physical chemistry. B, 117(4), 1186-1195 (2013-01-11)
High-resolution inelastic X-ray scattering was used to investigate the collective vibrational excitations in hydrated lysozyme powders as a function of hydration level and temperature. It is found that the samples with strong enzymatic function are "soft", in the sense that

Protocols

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service