Skip to Content
Merck
All Photos(1)

Key Documents

L7002

Sigma-Aldrich

L-Lysine p-nitroanilide dihydrobromide

≥98% (TLC), suitable for ligand binding assays

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N4O3 · 2HBr
CAS Number:
Molecular Weight:
428.12
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Lysine p-nitroanilide dihydrobromide,

Assay

≥98% (TLC)

Quality Level

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

storage temp.

2-8°C

SMILES string

Br.NCCCC[C@H](N)C(=O)Nc1ccc(cc1)N(=O)=O

InChI

1S/C12H18N4O3.BrH/c13-8-2-1-3-11(14)12(17)15-9-4-6-10(7-5-9)16(18)19;/h4-7,11H,1-3,8,13-14H2,(H,15,17);1H/t11-;/m0./s1

InChI key

FJNCJWJSSUJZMS-MERQFXBCSA-N

Application

L-Lysine p-nitroanilide may be used as a substrate to study the specificity and kinetics of lysine aminopeptidase(s) and various proteinases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natasa Bozić et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 134(2), 231-241 (2003-02-06)
Exopeptidases of Morimus funereus larvae were partially purified and characterized. Specific leucyl aminopeptidase (LAP) activity was increased eight-fold by gel filtration of the crude midgut extract. The partially purified LAP had a molecular mass greater than 100 kDa with pH
Akifumi Kawamura et al.
Biomacromolecules, 6(2), 627-631 (2005-03-15)
The amidase activity of bovine pancreas trypsin in water-soluble complexes with poly(ethylene glycol)-block-poly(alpha,beta-aspartic acid) (PEG-PAA) was evaluated by a colorimetric assay using L-lysine p-nitroanilide as a substrate. The enzymatic reaction of trypsin was accelerated through the complexation with PEG-PAA. By
Bernardo Ramírez-Zavala et al.
FEMS microbiology letters, 235(2), 369-375 (2004-06-09)
A lysine aminopeptidase was purified from the yeast Kluyveromyces marxianus. This enzyme was purified 100-fold from a soluble extract obtained at 100,000g. The purification procedure consisted in fractionated precipitation with ammonium sulfate and five chromatography steps. The native enzyme had
Priscilla L Phillips et al.
PloS one, 13(11), e0207295-e0207295 (2018-11-13)
The oral obligate anaerobe Porphyromonas gingivalis possesses a small conserved transcript PG_RS02100 of unknown function we previously identified using small RNA-seq analysis as expressed during logarithmic growth. In this study, we sought to determine if PG_RS02100 plays a role in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service