Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Product Name
D-Luciferin sodium salt, lyophilized powder, synthetic
biological source
synthetic
Quality Segment
assay
≥98% (HPLC)
form
lyophilized powder
solubility
H2O: soluble
fluorescence
λex 328 nm; λem 532 nm in H2O
storage temp.
−20°C
SMILES string
[Na+].Oc1ccc2nc(sc2c1)C3=N[C@H](CS3)C([O-])=O
InChI
1S/C11H8N2O3S2.Na/c14-5-1-2-6-8(3-5)18-10(12-6)9-13-7(4-17-9)11(15)16;/h1-3,7,14H,4H2,(H,15,16);/q;+1/p-1/t7-;/m1./s1
InChI key
LILQLBIQROYWIA-OGFXRTJISA-M
General description
Application
1 of 1
This Item | |||
|---|---|---|---|
| description lyophilized powder, synthetic | description synthetic, BioXtra, ≥99% (HPLC) | description synthetic | description ≥98.0% (HPLC) |
| assay ≥98% (HPLC) | assay ≥99% (HPLC) | assay ≥98% (HPLC) | assay ≥98.0% (HPLC) |
| solubility H2O: soluble | solubility DMSO: 10 mg/mL, clear, colorless to faintly yellow | solubility DMSO: 10 mg/mL, clear, colorless to faintly yellow | solubility water: soluble |
| Quality Level 200 | Quality Level 200 | Quality Level 300 | Quality Level 100 |
| form lyophilized powder | form powder | form powder | form powder or crystals |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| fluorescence λex 328 nm; λem 532 nm in H2O | fluorescence - | fluorescence λex 328 nm; λem 532 nm in H2O | fluorescence - |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


