Skip to Content
Merck
All Photos(1)

Key Documents

L5545

Sigma-Aldrich

Levosimendan

≥98% (HPLC)

Synonym(s):

(-)-OR-1259, (R)-Simendan, 2-[2-[4-[(4R)-1,4,5,6-Tetrahydro-4-methyl-6-oxo-3-pyridazinyl]phenyl]hydrazinylidene]-propanedinitrile, OR 1259, R)-((4-(1,4,5,6-Tetrahydro-4-methyl-6-oxo-3-pyridazinyl)phenyl)hydrazono) propanedintrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N6O
CAS Number:
Molecular Weight:
280.28
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -500 to -650°, c = 0.5 in THF

color

yellow

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

C[C@@H]1CC(=O)NN=C1c2ccc(N\N=C(\C#N)C#N)cc2

InChI

1S/C14H12N6O/c1-9-6-13(21)19-20-14(9)10-2-4-11(5-3-10)17-18-12(7-15)8-16/h2-5,9,17H,6H2,1H3,(H,19,21)/t9-/m1/s1

InChI key

WHXMKTBCFHIYNQ-SECBINFHSA-N

Application

Levosimendan has been used for screening its anti-human immunodeficiency virus type 1 (HIV-1) property. It has also been used for screening its cytotoxic effects in TP53 mutant and wild-type lung adenocarcinoma cell lines

Biochem/physiol Actions

Levosimendan has a potential to inhibit both acute human immunodeficiency virus type 1 (HIV-1) replication and the reactivation of latent HIV-1 proviruses. Therefore, it is considered to be a promising anti-HIV-1 agent.
Levosimendan is a calcium sensitiser.
Levosimendan is a calcium sensitiser. The compound increases the sensitivity of the heart to calcium, thus increasing cardiac contractility without a rise in intracellular calcium. Levosimendan binds to cardiac troponin C in a calcium-dependent manner and stabilizes troponin C. This is followed by actin-myosin cross-bridges, without increasing myocardial consumption of ATP. Additionally the compound acts as vasodilator by opening an ATP-sensitive potassium channels. Levosimendan is a drug used for treatment of congestive heart failure. Also it can be used to treat calcium channel blocker overdose.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saraswati Kache et al.
Pediatric research, 88(5), 705-716 (2020-07-08)
Fewer children than adults have been affected by the COVID-19 pandemic, and the clinical manifestations are distinct from those of adults. Some children particularly those with acute or chronic co-morbidities are likely to develop critical illness. Recently, a multisystem inflammatory
Claes-Håkan Bergh et al.
European journal of heart failure, 12(4), 404-410 (2010-03-26)
The aim of this study is to compare the effects of a 24 h intravenous infusion of levosimendan and a 48 h infusion of dobutamine on invasive haemodynamics in patients with acutely decompensated chronic NYHA class III-IV heart failure. All
Matti Adam et al.
Scientific reports, 5, 9704-9704 (2015-04-14)
Treatment of decompensated heart failure often includes administration of levosimendan. Myeloperoxidase (MPO) is released during polymorphonuclear neutrophil (PMN) degranulation, and mediates dysregulation of vascular tone in heart failure. We evaluated the effects of levosimendan-treatment on MPO in patients with acute
Malin Johansson et al.
Journal of cardiothoracic and vascular anesthesia, 28(4), 960-965 (2013-12-10)
The aim of the present study was to evaluate acute kidney injury (AKI) with cystatin C following transcatheter aortic valve implantation (TAVI) and to assess the impact of postoperative AKI on outcome and late renal function. A prospective study. Single
Enrique Guilherme et al.
Critical care (London, England), 24(1), 442-442 (2020-07-18)
Veno-arterial extracorporeal membrane oxygenation (VA-ECMO) has been increasingly used over the last decade in patients with refractory cardiogenic shock. ECMO weaning can, however, be challenging and lead to circulatory failure and death. Recent data suggest a potential benefit of levosimendan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service