Skip to Content
Merck
All Photos(2)

Documents

L5143

Sigma-Aldrich

Levorphanol (+)-tartrate salt dihydrate

white, powder, ≥98% (HPLC)

Synonym(s):

L-3-Hydroxy-N-methylmorphinan (+)-tartrate salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO · C4H6O6 · 2H2O
CAS Number:
Molecular Weight:
443.49
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

storage condition

protect from light

color

white

solubility

H2O: >20 mg/mL

InChI

1S/C17H23NO.C4H6O6.2H2O/c1-18-9-8-17-7-3-2-4-14(17)16(18)10-12-5-6-13(19)11-15(12)17;5-1(3(7)8)2(6)4(9)10;;/h5-6,11,14,16,19H,2-4,7-10H2,1H3;1-2,5-6H,(H,7,8)(H,9,10);2*1H2/t14-,16+,17+;1-,2-;;/m01../s1

InChI key

UMZNDVASJKIQCB-QLFXFZCRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Synthetic morphine analog that is a narcotic analgesic and μ opioid receptor agonist.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Morgan et al.
The Journal of pharmacology and experimental therapeutics, 289(2), 965-975 (1999-04-24)
Effects of low (butorphanol, nalbuphine)-, intermediate (buprenorphine)-, and high (morphine, levorphanol)-efficacy mu opioids were examined in F344, Sprague-Dawley (SD), Long-Evans (LE), and Lewis rats using a tail withdrawal and a drug discrimination procedure. In the tail withdrawal procedure using low
J R Bunzow et al.
Journal of neurochemistry, 64(1), 14-24 (1995-01-01)
We have cloned and expressed a rat brain cDNA, TS11, that encodes a mu-opioid receptor based on pharmacological, physiological, and anatomical criteria. Membranes were prepared from COS-7 cells transiently expressing TS11 bound [3H]diprenorphine with high affinity (KD = 0.23 +/-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service