Skip to Content
Merck
All Photos(1)

Key Documents

L4144

Sigma-Aldrich

Lectin from Phaseolus vulgaris (red kidney bean)

Leucoagglutinin PHA-L, lyophilized powder, BioReagent, suitable for cell culture

Synonym(s):

Phaseolus vulgaris agglutinin, PHA

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

product line

BioReagent

form

lyophilized powder

potency

erythroagglutination activity (Does not agglutinate a 2% suspension of human erythrocytes at 250 μg lectin/ml.)
≤1.0 μg per mL leucoagglutination activity
≤5 μg per mL mitogenic activity (Bromdeoxyuridine incorporation in human peripheral blood mononuclear cells)

technique(s)

cell culture | mammalian: suitable

impurities

salt, free

solubility

DPBS: 5 mg/mL, slightly hazy, colorless

suitability

leucocyte agglutination activity tested

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Lectins are carbohydrate-binding proteins,[1] which contains four subunits with a molecular weight of 125 kDa.[2] Leucoagglutinin PHA-L is a plant lectin, which is transported by the neurons in the central nervous system.[3]

Application

Lectin from Phaseolus vulgaris (red kidney bean) has been used:
  • it has been used as a positive control to treat cells[4]
  • to stimulate cell culture for the analysis of lymphocyte proliferation[5]
  • to stimulate T cells[6]

Biochem/physiol Actions

Lectin agglutinates cells and precipitates polysaccharides and glycoproteins.[1] It is associated with toxic allergens and hemagglutinins. Lectin exhibits anti-cancer, anti-human immunodeficiency virus (anti-HIV) and anti-microbial infection properties. Lectin inhibits mucosal atrophy, lowers type 2 diabetes and obesity, enhances nutrient absorption and regulates drug targeting.[7]
PHA consists of two molecular species, an erythroagglutinin (PHA-E) which has low mitogenic activity and high erythroagglutinin activity, and leucoagglutinin (PHA-L) which has high mitogenic and leucoagglutinating activity, but very low erythroagglutinating activity.
PHA-E is not blood group specific, but agglutination can be inhibited by certain oligosaccharides. PHA-P is the protein form of PHA prior to separation and purification of erythroagglutinin and leucoagglutinin. PHA-M is the mucoprotein form. Conjugates are prepared from the corresponding purified lectins.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a suspension of either human erythrocytes (2% in phosphate buffered saline, pH 6.8) or human leukocytes (107 per mL in saline) after 1 hr incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haifeng Huang et al.
Acta pharmaceutica Sinica. B, 10(7), 1321-1330 (2020-09-03)
JS001 (toripalimab) is a humanized IgG monoclonal antibody which strongly inhibits programmed cell death protein 1 (PD1). In this study, we used a different iodine isotype (nat/124/125I) to label JS001 probes to target the human PD1 (hPD1) antigen. In vitro
Christopher Lai-Hipp et al.
Transfusion, 51(2), 333-337 (2010-08-28)
Cellular substrate (CS) composed of phytohemagglutinin (PHA)-stimulated PBMNCs is required for isolating primary human immunodeficiency virus Type 1 from plasma (PHIV) during early acute infection. The transmitted founder PHIV is neutralization-sensitive and uses chemokine-receptor 5 to infect CD4-positive PBMNCs. Therefore
Transfection of FcgammaRIIIa (CD16) Alone Can Be Sufficient To Enable Human alphabetaTCR T Lymphocytes To Mediate Antibody-Dependent Cellular Cytotoxicity
Ollier J, et al.
ImmunoHorizons, 1(5), 63-70 (2017)
Novel DOCK8 gene mutations lead to absence of protein expression in patients with hyper-IgE syndrome
Qin T, et al.
Immunologic Research, 64(1), 260-271 (2016)
The Lectins: Properties, Functions, and Applications in Biology and Medicine (2012)

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Questions

  1. What do the designations -P, -L, -M, and -E represent in the names of PHA products?

    1 answer
    1. Generally, PHA-P or PHA-L is utilized to stimulate lymphocytes, although PHA-M will also work, but not to the same degree. The most active form is PHA-L (the mitogenic concentration is < 5μg/ml). PHA, or Phytohemagglutinin, comprises two molecular species:

      1. Erythroagglutinin (PHA-E), which demonstrates low mitogenic activity and high erythroagglutinating activity.

      2. Leucoagglutinin (PHA-L), which possesses high mitogenic and leucoagglutinating activity, accompanied by very low erythroagglutinating activity.

      PHA-P represents the protein form of PHA before the erythroagglutinin and leucoagglutinin components are separated and purified. PHA-M refers to the mucoprotein form of Phytohemagglutinin.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service