Skip to Content
Merck
All Photos(2)

Key Documents

L4025

Sigma-Aldrich

Lyticase from Arthrobacter luteus

lyophilized powder, ≥200 units/mg solid

Synonym(s):

(1,3)-β-D-Glucan endohydrolase, 1,3-β-Glucan glucohydrolase, Bacterial lyticase, Lysing enzyme

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bacterial (Arthrobacter luteus)

Quality Level

form

lyophilized powder

specific activity

≥200 units/mg solid

technique(s)

cell based assay: suitable

suitability

suitable for cell lysis

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Lyticase from Arthrobacter luteus has been used for resuspending cells to extract high-molecular weight DNA for polymerase chain reaction (PCR) fingerprint analyses.
Lyticase from Arthrobacter luteus has been used:
  • as a component in spheroplasting buffer to prepare yeast nuclear extracts[1]
  • for digestion in sample preparation for β-glucan enzymatic assay using yeast[1]
  • to lyse cells for RNA isolation[2]

Biochem/physiol Actions

Lyticase enzyme is frequently used in fungal research, particularly for species identification using polymerase chain reaction (PCR)-based techniques. It can break down β (1→3) and β (1→4) bonds between glucose units.
Lyticase hydrolyzes poly-β(1→3)-glucose such as yeast cell wall glucan.
Lyticase is a lysing enzyme that is used to extract DNA from yeast cells by inducing partial spheroplast formation.[3] Spheroplasts are subsequently lysed to release DNA. Lyticase is preferred to digest cell walls of yeast, which are difficult to disrupt because the cell walls may form capsules or resistant spores. Lyticase contains β-(1→;3)-glucan laminaripentaohydrolase along with β-(1→3)-glucanase, protease, and mannanase activities.[4] It is useful with yeast cells such as e.g. Candida,[5] Debaryomyces, Saccharomyces, Saccharomycopsis, Saccharomycodes, Eremothecium, and Schwanniomyces species.

Unit Definition

One unit will produce a ΔA800 of 0.001 per min at pH 7.5 at 25 °C, using a suspension of yeast as substrate in a 3 mL reaction mixture.

Other Notes

For R&D use only. Not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A protein transformation protocol for introducing yeast prion particles into yeast
Tanaka M, et al.
Methods in Enzymology, 470, 681-693 (2010)
M Groenewald et al.
Persoonia, 21, 17-27 (2008-12-01)
The type species of the genus Debaryomyces, Debaryomyces hansenii, is a highly heterogeneous species. It has been isolated from a large diversity of natural sources including fruit, air, water, soil, but most frequently from processed food products. The species delineation
Naturally occurring novel promoters around pyruvate branch-point for recombinant protein production in Pichia pastoris (Komagataella phaffii): pyruvate decarboxylase-and pyruvate kinase-promoters
Massahi A and Calik P
Biochemical Engineering Journal, 138, 111-120 (2018)
Alba Espargaró et al.
Microbial cell factories, 11, 89-89 (2012-06-27)
Prions were first identified as infectious proteins associated with fatal brain diseases in mammals. However, fungal prions behave as epigenetic regulators that can alter a range of cellular processes. These proteins propagate as self-perpetuating amyloid aggregates being an example of
Lisbeth Rosager Poulsen et al.
The Plant cell, 20(3), 658-676 (2008-03-18)
Vesicle budding in eukaryotes depends on the activity of lipid translocases (P(4)-ATPases) that have been implicated in generating lipid asymmetry between the two leaflets of the membrane and in inducing membrane curvature. We show that Aminophospholipid ATPase3 (ALA3), a member

Protocols

This procedure may be used for the determination of Lyticase activity using Baker’s yeast as the substrate.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service