Skip to Content
Merck
All Photos(4)

Key Documents

L3507

Sigma-Aldrich

Lignoceryl alcohol

≥99% (capillary GC)

Synonym(s):

1-Tetracosanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)23OH
CAS Number:
Molecular Weight:
354.65
Beilstein:
1777129
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99% (capillary GC)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCO

InChI

1S/C24H50O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25/h25H,2-24H2,1H3

InChI key

TYWMIZZBOVGFOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

424.4 °F - open cup

Flash Point(C)

218 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carminna Ottone et al.
Applied microbiology and biotechnology, 102(1), 237-247 (2017-11-02)
This work reports on the oxidation of long-chain aliphatic alcohols catalyzed by a stabilized alcohol dehydrogenase from S. cerevisiae (yeast alcohol dehydrogenase (YADH)). In particular, the oxidation of the fatty alcohol tetracosanol (C
Marion Callau et al.
Food chemistry, 333, 127403-127403 (2020-07-13)
Oil foams that are based on oleogels are stabilized by the presence of crystalline particles at the air bubble surface and in bulk. The size of crystalline particles is an important parameter in oil foam stabilization. The creation process (cooling
Donporn Wongwaiwech et al.
Scientific reports, 10(1), 21007-21007 (2020-12-05)
The byproducts of rice bran oil processes are a good source of fat-soluble nutraceuticals, including γ-oryzanol, phytosterol, and policosanols. This study aimed to investigate the effects of green technology with low pressure as the subcritical fluid extraction with dimethyl ether
Odimar Z Zanardi et al.
Scientific reports, 8(1), 455-455 (2018-01-13)
Under laboratory conditions, mating activity in Asian citrus psyllid (ACP) started 4 days after emergence, peaked at day 7, and showed a clear window of activity starting 8 h into the photophase and extending through the first hour of the scotophase.
Muthusamy Muthusamy et al.
Plants (Basel, Switzerland), 9(11) (2020-11-12)
The spectral quality and intensity of light, photoperiodism, and other environmental factors have profound impacts on the metabolic composition of light-dependent higher plants. Hence, we investigate the effects of fluorescent light (96 μmol m-2s-1) and white (100 μmol m-2s-1), blue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service