K3268
PAK7, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
KIAA1264, MGC26232, PAK5
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
47-63 nmol/min·mg
mol wt
~130 kDa
UniProt accession no.
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... PAK7(57144)
Biochem/physiol Actions
PAK7 (p21 protein (Cdc42/Rac)-activated kinase 7) belongs to the group-2 PAKs and can activate JNK (Jun N-terminus kinase). It contain an amino-terminal CDC42 (cell division cycle 42)/RAC (AKT1, v-akt murine thymoma viral oncogene homolog 1) interactive domain and a carboxyl terminal kinase domain. It plays an important role in outgrowth of neuron cells and synaptic vesicle trafficking. In mice, absence of PAK7 and PAK6 causes learning and locomotion defects. It is mainly expressed in brain. However, it is also considered as an oncogene because of its expression in colorectal, esophageal and gastric cancers.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
PloS one, 9(12), e113989-e113989 (2014-12-02)
Aurora-A overexpression is common in various types of cancers and has been shown to be involved in tumorigenesis through different signaling pathways, yet how the deregulation affects cancer therapeutics remains elusive. Here we showed that overexpression of Aurora-A rendered esophageal
Molecular and cellular biochemistry, 383(1-2), 191-199 (2013-07-24)
P21-activated kinase 5 (PAK5) is the recently identified member of the group II p21-activated kinases (PAKs) family, which is characterized by a highly conserved amino-terminal Cdc42/Rac interactive binding domain and a carboxyl terminal kinase domain. However, the role of PAK5
Cell biology international, 38(12), 1394-1402 (2014-07-24)
p21-activated kinase 7 (PAK7), also named as PAK5, is a member of Rac/Cdc42-associated Ser/Thr protein kinases. It is overexpressed in some types of cancer such as colorectal and pancreatic cancers. However, the expression status and biological function of PAK7 in
Oncogene, 21(24), 3939-3948 (2002-05-29)
The p21-activated kinase (PAK) family of protein kinases has recently attracted considerable attention as an effector of Rho family of small G proteins and as an upstream regulator of MAPK signalling pathways during cellular events such as re-arrangement of the
The international journal of biochemistry & cell biology, 34(7), 713-717 (2002-04-16)
The p21-activated kinases (Paks) are serine/threonine protein kinases that bind to and, in some cases, are stimulated by activated forms of the small GTPases, Cdc42 and Rac. With the recent discovery of several novel isoforms, Paks are now categorized into
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service