Skip to Content
Merck
All Photos(1)

Key Documents

I7267

Sigma-Aldrich

Insect Medium Supplement (10×)

liquid, sterile-filtered, BioReagent, suitable for insect cell culture

Synonym(s):

10× Insect Supplement, Insect Medium, Medium Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

sterility

sterile-filtered

product line

BioReagent

form

liquid

technique(s)

cell culture | insect: suitable

storage temp.

2-8°C

Application

A serum replacement for general cell growth and maintenance.
May be used in the preparation of low protein medium. In many instances the level of serum can be reduced by 50-75% in cultures containing 10% medium supplement. The low protein content (<150 μg/ml) also makes the product suitable for use in preparing low protein serum-free medium. Serum-free medium can be prepared by adding the medium supplement to a final concentration of 10% to an appropriate cell culture medium. The final protein content of the medium will be dependent upon the level of proteins (if any) present in the basal medium. In many instances the adaptation of cells to growth in serum-free medium may be required. Also recommended for use with serum to improve cell growth and maintenance.

Other Notes

Proprietary mixture of amino acids, lipids, trace elements, vitamins and other growth enhancing components in an aqueous solution.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Mische et al.
Molecular biology of the cell, 19(11), 4918-4929 (2008-09-19)
The dynein light intermediate chain (LIC) is a subunit unique to the cytoplasmic form of dynein, but how it contributes to dynein function is not fully understood. Previous work has established that the LIC homodimer binds directly to the dynein
Jeong-Hoon Han et al.
Cell death & disease, 11(11), 944-944 (2020-11-04)
Wg/Wnt as a signaling protein binds to Frizzled (Fz) and Arrow (Arr), two Wg co-receptors essential for Wg signaling for cell proliferation, differentiation, and cell survival. Arr has a long extracellular region, a single transmembrane domain and an intracellular region.
Dong-Gyu Cho et al.
Molecules and cells, 44(1), 13-25 (2021-01-30)
Apoptosis and compensatory proliferation, two intertwined cellular processes essential for both development and adult homeostasis, are often initiated by the mis-regulation of centrosomal proteins, damaged DNA, and defects in mitosis. Fly Anastral spindle 3 (Ana3) is a member of the
Emily Tubman et al.
Cellular and molecular bioengineering, 11(1), 25-36 (2018-03-20)
The microtubule motor protein kinesin-5 is well known to establish the bipolar spindle by outward sliding of antiparallel interpolar microtubules. In yeast, kinesin-5 also facilitates chromosome alignment "congression" at the spindle equator by preferentially depolymerizing long kinetochore microtubules (kMTs). The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service