Skip to Content
Merck
All Photos(1)

Documents

I6505

Sigma-Aldrich

Diethylaminoethyl–Sephacel®

aqueous ethanol suspension, 40-160 μm (wet), exclusion limit ~1,000,000 Da

Synonym(s):

DEAE-Sephacel®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47101511
PubChem Substance ID:
NACRES:
NA.56

form

aqueous ethanol suspension

technique(s)

activity assay: suitable

particle size

40-160 μm (wet)

pore size

~1,000,000 Da exclusion limit

pH

2—12

capacity

100-140 μeq/mL, gel

storage temp.

2-8°C

SMILES string

OC[C@@H]1O[C@@H](O[C@H]2[C@@H](O)[C@H](O)[C@@H](O)O[C@H]2CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

General description

I6505-500ML′s update product number is GE17-0500-01

Application

Diethylaminoethyl (DEAE) Sephacel® is used in protein chromatography, ion exchange chromatography, and anion exchange media. It has been used to study sperm motility, sperm capacitation and sperm-oocyte interactions. DEAE Sephacel has also been used to study patients with immunoglobulin A (IgA) nephropathy. Diethylaminoethyl–Sephacel® has been used:

  • in the purification of media samples by ion-exchange chromatography
  • in the isolation of glycosaminoglycans (GAG) from cultured animal cells
  • in the purification of cytotoxic fractions from Tritrichomonas foetus isolates

Physical form

Suspension in 20% ethanol

Legal Information

Sephacel is a registered trademark of Cytiva

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

129.2 °F

Flash Point(C)

54 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H A Frank et al.
Photosynthesis research, 37(3), 193-203 (1993-09-01)
Triplet state electron paramagnetic resonance (EPR) experiments have been carried out at X-band on Rb. sphaeroides R-26 reaction centers that have been reconstituted with the carotenoid, spheroidene, and exchanged with 13(2)-OH-Zn-bacteriochlorophyll a and [3-vinyl]-13(2)-OH-bacteriochlorophyll a at the monomeric, 'accessory' bacteriochlorophyll
C M Rembold et al.
Biochimica et biophysica acta, 1500(3), 257-264 (2000-03-04)
Nitrovasodilators, high extracellular Mg(2+), and some other relaxing agents can cause smooth muscle relaxation without reductions in myosin regulatory light chain (MRLC) phosphorylation. Relaxations without MRLC dephosphorylation suggest that other regulatory systems, beyond MRLC phosphorylation, are present in smooth muscle.
Daniel R Studelska et al.
Glycobiology, 16(1), 65-72 (2005-09-17)
Glycosaminoglycans (GAGs) are linear polysaccharides made by all animal cells. GAGs bind to hundreds of proteins, such as growth factors, cytokines, chemokines, extracellular matrix components, protease inhibitors, proteases, and lipoprotein lipase, through carbohydrate and protein interactions. These interactions control many
Pierina De Muro et al.
Scandinavian journal of urology and nephrology, 41(3), 230-236 (2007-05-01)
A 2-year follow-up study was carried out in patients with IgA nephropathy (IgAN) in order to verify the possible use of quali-quantitative analysis of urinary glycosaminoglycans (GAGs) as a prognostic index of disease and for drug treatment monitoring. Ten patients
V T D'Souza et al.
Biochemical and biophysical research communications, 129(3), 727-732 (1985-06-28)
An artificial chymotrypsin, with all the features of the real chymotrypsin, namely a binding site (from cyclodextrin) attached to a catalytic site containing an imidazolyl group, a carboxylate group and a hydroxyl group, has been synthesized. This artificial chymotrypsin has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service