Skip to Content
Merck
All Photos(1)

Key Documents

I5152

Sigma-Aldrich

Anti-Interferon-γ Receptor 1 (IFN-γ R1) antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-CD119, Anti-IFNGR, Anti-IMD27A, Anti-IMD27B

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

immunocytochemistry: 5-15 μg/mL using immersion fixed human peripheral blood mononuclear cells
neutralization: suitable
western blot: 0.1 μg/mL

isotype

IgG

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFNGR1(3459)

General description

Human Interferon-γ- Receptor I (IFN-γ RI) is a 25kD, 227 amino acid residue containing protein that belongs to type II cytokine receptor family. IFN-γ RI has a crucial role in ligand-binding and signaling. Ligand IFN-γ induces tyrosine phosphorylation in IFN-γ RI which in turn leads to the formation of a docking site on the activated receptor for Stat1 that specifically activates IFN-γ induced gene transcription. Anti-Interferon γ Receptor I (IFN-γ RI) antibody can be used in neutralization assay using a confluent culture of HeLa cells. Goat anti-Interferon γ Receptor I (IFN-γ RI) antibody reacts specifically with recombinant human IFN-γ RI. The product has shown no cross-reactivity for recombinant human IFN-γ RII.

Immunogen

recombinant human IFN-γ Receptor I extracellular domain expressed in NSO cells.

Application

Anti-Interferon γ Receptor I (IFN-γ RI) antibody can be used in indirect ELISA and western blot.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Silvennoinen et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 105(7), 497-509 (1997-07-01)
Cytokines are the principal regulators of cell proliferation and differentiation of hematopoietic cells and these responses are initiated through activation of hematopoietic cytokine receptors. Although the receptor intracellular domains lack any kinase domains, activation of cytokine receptors lead to rapid
E A Bach et al.
Annual review of immunology, 15, 563-591 (1997-01-01)
During the last several years, the mechanism of IFN gamma-dependent signal transduction has been the focus of intense investigation. This research has recently culminated in the elucidation of a comprehensive molecular understanding of the events that underlie IFN gamma-induced cellular
A C Greenlund et al.
The EMBO journal, 13(7), 1591-1600 (1994-04-01)
Herein we report that interferon-gamma (IFN gamma) induces the rapid and reversible tyrosine phosphorylation of the IFN gamma receptor. Using a panel of receptor intracellular domain mutants, we show that a membrane-proximal LPKS sequence (residues 266-269) is required for ligand-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service