Skip to Content
Merck
All Photos(1)

Key Documents

I3525

Sigma-Aldrich

Interleukin-17A human

≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

CTLA-8, IL-17

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≤2.0 ng/mL ED50

mol wt

31 kDa

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested (endotoxin level is less than 0.1 ng per μg cytokine (1 EU/μg).)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL17A(3605)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

IL-17, initially known as CTLA-8, is a T cell-expressed polypeptide that appears to be involved in the regulation of the hematopoietic system. IL-17 has been shown to induce IL-6, IL-8, and G-CSF production by fibroblasts; all of these cytokines have demonstrated effects on hematopoiesis.

Physical form

Lyophilized with no additives.

Analysis Note

  • The biological activity of human IL-17 is measured by its ability to induce IL-6 production using NHDF cells.
  • Biological activity is also determined by the dose-dependent induction of IL-6 in primary human foreskin fibroblast. The expected ED50 is < 2.0 ng/ml, corresponding to a specific activity of > 5 x 105 units/mg.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yan Jin et al.
The Journal of asthma : official journal of the Association for the Care of Asthma, 52(10), 981-988 (2015-08-25)
There has been significant interest in the association between asthma and the polymorphisms of IL-17A and IL-17F for a period of time. This work aims to present a clearer relationship between asthma and the polymorphisms of IL-17A and IL-17F. Searches
Lin Fan et al.
PloS one, 10(5), e0127087-e0127087 (2015-05-29)
Th1 cells play an essential role in immune protection against tuberculosis. Th17 cells might be involved with immune pathology in active human tuberculosis (TB). The balance between Th1 and Th17 cells in patients with cavitary tuberculosis needs to be clarified
W T Qi et al.
Genetics and molecular research : GMR, 14(4), 13364-13369 (2015-11-05)
We conducted a study to investigate the role of three IL-17 gene single nucleotide polymorphisms (SNP) (rs2275913G>A, rs3748067C>T, and rs763780 T>C) in the development of gastric cancer. A total of 252 patients with gastric cancer and 252 control subjects were
J Y Wang et al.
Allergy, 64(7), 1056-1060 (2009-02-13)
The interleukin 17A (IL17A) gene, located on chromosome 6p and linked to asthma phenotype, is a highly potential candidate gene conferring asthma susceptibility. The purpose of this study was to investigate the genetic association between single nucleotide polymorphisms (SNPs) of
Junhua Wang et al.
PLoS neglected tropical diseases, 9(5), e0003755-e0003755 (2015-05-09)
The growth potential of the tumor-like Echinococcus multilocularis metacestode (causing alveolar echinococcosis, AE) is directly linked to the nature/function of the periparasitic host immune-mediated processes. We previously showed that Fibrinogen-like-protein 2 (FGL2), a novel CD4+CD25+ Treg effector molecule, was over-expressed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service