Skip to Content
Merck
All Photos(1)

Key Documents

I1630

Sigma-Aldrich

Intestinal acetone powders from rat

Synonym(s):

Rattus rattus intestine acetone powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

biological source

rat

Quality Level

form

powder

storage temp.

−20°C

Application

Intestinal acetone powder rat has been used:
  • to degrade salicin-α-D-maltotriopyranoside (β-Sa-α-G3)
  • as a reagent for the evaluation of α-glucosidase inhibition
  • in a study to obtain angiotensin I converting enzyme (ACE) inhibitory active fractions from pea and whey protein hydrolysates

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of probiotic potential and anti-hyperglycemic properties of a novel Lactobacillus strain isolated from water kefir grains
Koh WY, et al.
Food Science and Biotechnology, 27(5), 1369-1376 (2018)
Fractionation of angiotensin I converting enzyme inhibitory activity from pea and whey protein in vitro gastrointestinal digests
Vermeirssen V, et al.
Journal of the Science of Food and Agriculture, 85(3), 399-405 (2005)
Elizabeth Devi Wahengbam et al.
Food chemistry, 292, 39-46 (2019-05-06)
Iron (Fe) and folic acid (FA) fortified parboiled rice was produced by applying 'brown rice parboiling' method. The effect of milling and the effectiveness of fortification were tested in relation to the amount of bioaccessible and bioavailable form of Fe
Wee Yin Koh et al.
Food science and biotechnology, 27(5), 1369-1376 (2018-10-16)
A total of eight strains of lactic acid bacteria were isolated from water kefir grains and assessed for their in vitro α-glucosidase inhibitory activity. Lactobacillus mali K8 demonstrated significantly higher inhibition as compared to the other strains, thus was selected
Application of amylomaltase for the synthesis of salicin-alpha-glucosides as efficient anticoagulant and anti-inflammatory agents
Rudeekulthamrong P and Kaulpiboon J
Carbohydrate Research, 432, 55-61 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service