Skip to Content
Merck
All Photos(1)

Key Documents

HT501640

Sigma-Aldrich

Formalin solution, neutral buffered, 10%

histological tissue fixative

Synonym(s):

Formaldehyde, Methanal

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

concentration

10% formalin (approx. 4% formaldehyde)

color

colorless

pH

6.90-7.10

density

1.080 g/cm3

application(s)

hematology
histology

storage temp.

room temp

SMILES string

[O-]#[C+H2]

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Formalin solution, neutral buffered, 10% is the most commonly used fixative for light microscopy. It requires a relatively short period of fixation. It is suitable for the long-term storage of tissue. Once properly fixed, the tissue should be able to withstand the subsequent stages of tissue processing or staining. 10% NBF is well suited for large throughput laboratories, and requires a relatively short period of fixation, but can also be used for the long-term storage of tissue. Formalin fixation is thought to form between the aldehydes and the proteins, creating a gel, thus retaining cellular constituents in their in vivo relationship.

Application

Formalin solution, neutral buffered, 10% has been used as a popular general purpose histological fixative.[1][2]It helps to preserve tissue samples by preventing decay and maintaining their molecular integrity.
Popular general purpose histological fixative.

Packaging

Item is packaged in a 20 L Cubitainer consisting of a plastic bladder inside of a cardboard box with an attached spigot.

Physical form

Neutral buffered solution

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Muta. 2 - Skin Sens. 1

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fan Liu et al.
Journal of biomaterials applications, 33(9), 1168-1177 (2019-01-23)
To investigate the osteogenesis of macro-pore sized bone scaffolds, biphasic calcium phosphate scaffolds with accurately controlled macro-pore size (0.8, 1.2, and 1.6 mm) and identical porosity of 70% were fabricated by the 3D printing technology. Eight New Zealand rabbits were
B cell autophagy mediates TLR7-dependent autoimmunity and inflammation.
Weindel C G, et al
Autophagy, 11(5), 1010-1024 (2015)
Autophagy in Dendritic Cells and B Cells Is Critical for the Inflammatory State of TLR7-Mediated Autoimmunity.
Weindel C G, et al.
Journal of Immunology, 198, 1081-1092 (2016)
Ming-Kai Hsieh et al.
Scientific reports, 10(1), 10441-10441 (2020-06-28)
Screw loosening due to broken pedicles is a common complication resulting from the insertion of screws either with inadequate diameters or into an osteoporotic pedicle. In this novel in vitro study, we tried to clarify the contribution of the pedicle
Xiaozhen Liang et al.
PLoS pathogens, 7(9), e1002220-e1002220 (2011-09-21)
Human gammaherpesviruses are associated with the development of lymphoproliferative diseases and B cell lymphomas, particularly in immunosuppressed hosts. Understanding the molecular mechanisms by which human gammaherpesviruses cause disease is hampered by the lack of convenient small animal models to study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service