Skip to Content
Merck

HT102A

Reticulum Stain Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
UNSPSC Code:
41116121
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label

IVD

for in vitro diagnostic use

dilution

(for histology)

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

General description

The kit is based upon the method of Gordon and Sweets.

Application

Intended to demonstrate reticular fibers in paraffin sections.


Kit Components Only

Product No.
Description

  • Ferric Ammonium Sulfate Solution (kit only) 500 mL

  • Gold Chloride Solution (kit only) 100 mL

  • Oxalic Acid Solution (kit only) 100 mL

  • Potassium Permanganate Solution (kit only) 100 mL

  • Silver Nitrate Solution (kit only) 50 mL

  • Sodium Hydroxide Solution (kit only) 100 mL

  • Sodium Thiosulfate Solution (kit only) 500 mL

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library