About This Item
NACRES:
NB.22
UNSPSC Code:
41122103
sterility
sterile
feature
individually wrapped
packaging
pk of 500
manufacturer/tradename
Heathrow Scientific
size × width × thickness
172 mm × 37 mm × 4 mm
Features and Benefits
- Smooth spreading is aided by a domed headpiece.
- Agar tears are reduced with a flexible rippled handle and domed spreading bar.
- Disposable
- γ sterilized
Legal Information
Delta is a trademark of Heathrow Scientific LLC
Choose from one of the most recent versions:
Already Own This Product?