Skip to Content
Merck
All Photos(2)

Key Documents

HPA077249

Sigma-Aldrich

Anti-ETV1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

Synonym(s):

ER81

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

ASQSFPPPLMIKQEPRDFAYDSEVPSCHSIYMRQEGFLAHPSRTEGCMFEKGPRQFYDDTCVVPE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ETV1(2115)

General description

ETS translocation variant 1 (ETV1) is transcriptional activator and is expressed in two isoforms. It has a winged helix-turn-helix ETS DNA binding motif. ETV1 is mapped to human chromosome 7p21.2.

Immunogen

ets variant 1

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ETS translocation variant 1 (ETV1) plays a key role in signalling and development. Phosphorylation of ETV1 bymitogen- and stress-activated protein kinase 1 (MSK1) activates transcriptional regulation. ETV1 promotes tumor progression by triggering ETS transcription factors and is highly expressed gastrointestinal stromal tumours.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST90428

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of the ETS transcription factor ER81 by the 90-kDa ribosomal S6 kinase 1 and protein kinase A
Wu J and Janknecht R
The Journal of Biological Chemistry, 277(45), 42669-42679 (2002)
Characterization of the human and mouse ETV1/ER81 transcription factor genes: role of the two alternatively spliced isoforms in the human
Coutte L, et al.
Oncogene, 18(46), 6278-6286 (1999)
An oncogenic role for ETV1 in melanoma
Jane-Valbuena J, et al.
Cancer Research, 0008-5472 (2010)
ETV1 mRNA is specifically expressed in gastrointestinal stromal tumors
Jang BG, et al.
Virchows Archiv, 467(4), 393-403 (2015)
ETV1 is a lineage survival factor that cooperates with KIT in gastrointestinal stromal tumours
Chi P, et al.
Nature, 467(7317), 849-849 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service