Skip to Content
Merck
All Photos(3)

Key Documents

HPA038356

Sigma-Aldrich

Anti-PCBP2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HNRNPE2, Anti-HNRPE2, Anti-hnRNP-E2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

IFAGGQDRYSTGSDSASFPHTTPSMCLNPDLEG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCBP2(5094)

General description

Poly (rC) binding protein 2 (PCBP2), also known as heterogeneous ribonuclear protein E2 (hnRNP E2) and αCP2, belongs to a group of proteins that bind to poly(C) stretches of both RNA and DNA and is encoded by the gene mapped to human chromosome 12q13.13. The encoded protein is a key constituent of stress granules (SG) and processing bodies (P-body). The protein shuttles between cytoplasm and nucleus.

Immunogen

poly(rC) binding protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry. The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-PCBP2 antibody produced in rabbit has been used in fully automated and standardized-individual-nucleotide resolution cross-linking and immunoprecipitation (FAST- iCLIP).

Biochem/physiol Actions

Poly (rC) binding protein 2 (PCBP2) plays a vital role in controlling mRNA stability and regulating translation 14. In addition, it can also take part in protein-protein interactions. The encoded protein functions as a negative regulator of mitochondrial antiviral-signaling protein (MAVS)-mediated antiviral signaling via interacting with HECT (homologous to the E6-AP carboxyl terminus) domain -containing E3 ligase AIP4. Dysregulated expression of the gene results in oral cancer. PCBP2 plays a vital role as an initiator of internal ribosomal entry site (IRES)-mediated translation of both viral and cellular mRNA. Additionally, it also facilitates many biological processes, such as transcriptional regulation and translational silencing. Elevated expression of PCBP2 has been observed in human glioma tissues and cell lines. Downregulation of this gene, by the inhibition of cell-cycle progression and activation of caspase-3–mediated apoptosis, retards glioma growth in vitro and in vivo.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81245

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PCBP2 mediates degradation of the adaptor MAVS via the HECT ubiquitin ligase AIP4.
You F
Nature Immunology, 10, 1300-1308 (2009)
Ryan A Flynn et al.
RNA (New York, N.Y.), 21(1), 135-143 (2014-11-21)
RNA-protein interactions are central to biological regulation. Cross-linking immunoprecipitation (CLIP)-seq is a powerful tool for genome-wide interrogation of RNA-protein interactomes, but current CLIP methods are limited by challenging biochemical steps and fail to detect many classes of noncoding and nonhuman
Dissecting noncoding and pathogen RNA-protein interactomes.
Flynn RA
RNA, 21, 135-143 (2015)
Common Dysregulated Genes in Endometriosis and Malignancies.
Dentillo DB
Revista Brasileira de Ginecologia e obstetricia : Revista da Federac?o Brasileira das Sociedades de Ginecologia e Obstetricia, 38, 253-262 (2016)
Ken Fujimura et al.
RNA (New York, N.Y.), 14(3), 425-431 (2008-01-05)
Recent advances in microscopic techniques have shed light on the roles of specific subcellular structures in the regulation of gene expression. One such structure is the stress granule (SG), which is engaged in stress-triggered translational arrest by sequestering pre-initiation complexes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service