Skip to Content
Merck
All Photos(2)

Key Documents

HPA028417

Sigma-Aldrich

Anti-PHF20L1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2

Synonym(s):

Anti-CGI-72, Anti-FLJ13649, Anti-FLJ21615, Anti-MGC64923, Anti-TDRD20B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

EETSTCIATPDVEKKEDLPTSSETFGLHVENVPKMVFPQPESTLSNKRKNNQGNSFQAKRARLNKITGLLASKAVGVDGAEKKEDYNETA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHF20L1(51105)

General description

The gene PHF20L1 (PHD finger protein 20-like 1) is mapped to human chromosome 8q24.22. The protein has a MBT (malignant brain tumor) domain and tudor domain.

Immunogen

PHD finger protein 20-like 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-PHF20L1 antibody produced in rabbit has been used in western blotting and immunoprecipitation.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

PHF20L1 (PHD finger protein 20-like 1) interacts with monomethylated lysine 142 on DNMT1 (DNA methyltransferase 1). It helps chromatin loading of DNMT1 and also protects it from proteasomal degradation. PHF20L1 might play a crucial role in epigenetic inheritance. The gene is overexpressed in breast cancer and is linked with shorter survival of patients. Absence of PHF20L1 activity suppresses cell proliferation in breast cancer cell lines.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76232

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dulce Rosario Alberto-Aguilar et al.
Cancer cell international, 22(1), 6-6 (2022-01-08)
Ovarian cancer is the most aggressive gynecological malignancy. Transcriptional regulators impact the tumor phenotype and, consequently, clinical progression and response to therapy. PHD finger protein 20-like protein 1 (PHF20L1) is a transcriptional regulator with several isoforms, and studies on its
Chunxiao Zhang et al.
The Journal of biological chemistry, 294(2), 476-489 (2018-11-18)
SOX2 is a dose-dependent master stem cell protein that controls the self-renewal and pluripotency or multipotency of embryonic stem (ES) cells and many adult stem cells. We have previously found that SOX2 protein is monomethylated at lysine residues 42 and
Dulce Rosario Alberto-Aguilar et al.
Cancer microenvironment : official journal of the International Cancer Microenvironment Society, 12(2-3), 181-195 (2019-07-04)
Ovarian cancer is considered to be the most lethal type of gynecological cancer. During the advanced stages of ovarian cancer, an accumulation of ascites is observed. Fucosylation has been classified as an abnormal post-translational modification that is present in many
Simon M Carr et al.
Cell death and differentiation, 24(12), 2139-2149 (2017-08-26)
The retinoblastoma tumour suppressor protein (pRb) classically functions to regulate early cell cycle progression where it acts to enforce a number of checkpoints in response to cellular stress and DNA damage. Methylation at lysine (K) 810, which occurs within a
Feng Leng et al.
Nature communications, 9(1), 1641-1641 (2018-04-25)
Many non-histone proteins are lysine methylated and a novel function of this modification is to trigger the proteolysis of methylated proteins. Here, we report that the methylated lysine 142 of DNMT1, a major DNA methyltransferase that preserves epigenetic inheritance of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service