Skip to Content
Merck
All Photos(4)

Key Documents

HPA025926

Sigma-Aldrich

Anti-NMU antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Neuromedin-U, Anti-NmU-25

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

ILPQGLQPEQQLQLWNEIDDTCSSFLSIDSQPQASNALEELCFMIMGMLPKPQEQDEKDNTKRFLFHYSKTQKLGKSNVVSSVVHPLLQLVPHLHERRMKRFRVDEEFQSPFASQSRGYFLFRPRNGRRSAGFI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NMU(10874)

Related Categories

General description

Neuromedin U (NMU) is a hypothalamic neuropeptide. It is expressed in brain, adipose tissue and gastroenteric tract. NMU is located on human chromosome 4.

Immunogen

Neuromedin-U Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-NMU antibody has been used in immunohistochemical staining and western blotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Neuromedin U (NMU) plays a major role in various metabolic processes like appetite regulation, energy homeostasis, gastric secretion, smooth muscle contraction and bone remodeling. It also participates in the development of various cancers. NMU is considered as a therapeutic target for obesity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70217

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of neuromedin U in adiposity regulation. Haplotype analysis in European children from the IDEFICS Cohort.
Gianfagna F, et al.
PLoS ONE, 12(2), e0172698-e0172698 (2017)
Lei Wang et al.
Molecular medicine reports, 14(2), 1075-1082 (2016-06-10)
Regional metastasis is an important prognostic factor for patients with head and neck squamous cell carcinoma (HNSCC). Neuromedin U (Nmu) is a secreted neuropeptide, named due to its potent uterine contraction‑inducing activity. The aim of the present study was to
Overexpression of neuromedin U is correlated with regional metastasis of head and neck squamous cell carcinoma.
Wang L, et al.
Molecular Medicine Reports, 14(2), 1075-1082 (2016)
Oncogenic features of neuromedin U in breast cancer are associated with NMUR2 expression involving crosstalk with members of the WNT signaling pathway.
Garczyk S, et al.
Oncotarget, 8(22), 36246-36246 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service